Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821379_g1
          See other DEDD GE Assays ›
          Gene Symbol:
          Dedd
          Gene Name:
          death effector domain-containing
          Gene Aliases:
          Deft
          Chromosome Location:
          Chr.13: 89708477 - 89722482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821379_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dedd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031800.1 NP_113988.1 2 - 3 478 75
          XM_006250269.2 XP_006250331.1 4 - 5 543 75
          XM_006250270.2 XP_006250332.1 2 - 3 493 75
          XM_006250272.2 XP_006250334.1 4 - 5 623 75
          XM_006250273.2 XP_006250335.1 3 - 4 549 75
          XM_006250274.2 XP_006250336.1 2 - 3 497 75
          GenBank mRNA AF053362.1 - 2 - 3 478 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dedd

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          apoptotic process
          spermatogenesis
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of protein catabolic process
          regulation of apoptotic process
          decidualization
          negative regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P662710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1513 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P662710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P662710 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P662710 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P662710 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700696409 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100775 - -
          Agilent Rat cDNA Microarray (G4105A) 700696409 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765552 - -
          Affymetrix Rat Gene 1.0 ST Array 10765556 - -
          Affymetrix Rat Gene 1.0 ST Array 10765554 - -
          Affymetrix Rat Gene 1.0 ST Array 10765553 - -
          Affymetrix Rat Gene 1.1 ST Array 10765551 - -
          Rat Genome 230 2.0 Array 1382326_at - -
          Rat Genome 230 2.0 Array 1369003_PM_at - -
          Rat Genome 230 2.0 Array 1369003_at - -
          Rat Genome 230 2.0 Array 1382326_PM_at - -
          Affymetrix HT Rat Focus Array 1369003_at - -
          Affymetrix HT Rat Focus Array 1382326_PM_at - -
          Affymetrix HT Rat Focus Array 1369003_PM_at - -
          Affymetrix HT Rat Focus Array 1382326_at - -
          Affymetrix Rat Clariom S Assay PSR1300005276.rn.1 TC1300000758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005277.rn.1 TC1300000758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005275.rn.1 TC1300000758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300005278.rn.1 TC1300000758.rn.1 RTA TC
          Rat Expression Set 230 1369003_PM_at - -
          Rat Expression Set 230 1369003_at - -
          Rat Expression Set 230 1382326_at - -
          Rat Expression Set 230 1382326_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765551 - -
          Rat Genome U34 Set AF053362_g_at - -
          Rat Genome U34 Set AF053362_at - -
          Rat Genome U34 Set rc_AA850569_at - -
          Rat Genome U34 Set AF053362_g_at - -
          Rat Genome U34 Set AF053362_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22297875 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364350 - -
          RatRef-12 Expression BeadChip ILMN_58637 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline