Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821381_g1
          See other NME2 GE Assays ›
          Gene Symbol:
          Nme2
          Gene Name:
          NME/NM23 nucleoside diphosphate kinase 2
          Gene Aliases:
          NDKB, nm23-2, p18-12d
          Chromosome Location:
          Chr.10: 81648216 - 81653717 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821381_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nme2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031833.2 NP_114021.2 2 - 3 295 91
          GenBank mRNA BC086599.1 - 2 - 3 295 7312852 91
          FQ217120.1 - 1 - 2 196 91
          FQ217563.1 - 2 - 3 182 91
          FQ218179.1 - 1 - 2 212 91
          FQ222529.1 - 2 - 3 180 91
          M55331.1 - 1 - 2 152 91
          M91597.1 - 2 - 3 186 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nme2

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of myeloid leukocyte differentiation
          nucleoside diphosphate phosphorylation
          GTP biosynthetic process
          UTP biosynthetic process
          CTP biosynthetic process
          integrin-mediated signaling pathway
          nucleoside triphosphate biosynthetic process
          positive regulation of neuron projection development
          peptidyl-histidine phosphorylation
          positive regulation of cAMP biosynthetic process
          cellular response to oxidative stress
          regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of keratinocyte differentiation
          regulation of epidermis development
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          positive regulation of epithelial cell proliferation
          protein oligomerization
          response to growth hormone
          cellular response to glucose stimulus
          cellular response to fatty acid
          nucleoside diphosphate kinase activity
          protein histidine kinase activity
          protein serine/threonine kinase activity
          fatty acid binding
          ATP binding
          drug binding
          intermediate filament binding
          enzyme binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P130504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130504 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P130504 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P130504 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1040567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746272 - -
          Affymetrix Rat Gene 1.0 ST Array 10746274 - -
          Affymetrix Rat Gene 1.0 ST Array 10746273 - -
          Affymetrix Rat Gene 1.0 ST Array 10746275 - -
          Affymetrix Rat Gene 1.1 ST Array 10746271 - -
          Rat Genome 230 2.0 Array 1367766_PM_at - -
          Rat Genome 230 2.0 Array 1367766_at - -
          Rat Genome 230 2.0 Array 1391710_PM_at - -
          Rat Genome 230 2.0 Array 1391710_at - -
          Affymetrix HT Rat Focus Array 1367766_at - -
          Affymetrix HT Rat Focus Array 1367766_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000031799.rn.1 TC1000003575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031800.rn.1 TC1000003575.rn.1 RTA TC
          Rat Expression Set 230 1367766_PM_at - -
          Rat Expression Set 230 1367766_at - -
          Rat Expression Set 230 1391710_at - -
          Rat Expression Set 230 1391710_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746271 - -
          Rat Genome U34 Set M91597_s_at - -
          Rat Genome U34 Set M91597_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20822398 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370390 - -
          RatRef-12 Expression BeadChip ILMN_58720 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline