Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821407_g1
          See other CALM1 GE Assays ›
          Gene Symbol:
          Calm1
          Gene Name:
          calmodulin 1
          Gene Aliases:
          CaMI
          Chromosome Location:
          Chr.6: 124217241 - 124225292 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821407_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031969.2 NP_114175.1 5 - 6 534 82
          GenBank mRNA AF178845.1 - 469 82
          FQ211760.1 - 5 - 6 601 82
          FQ212721.1 - 939 82
          FQ213013.1 - 1 - 2 96 82
          FQ213227.1 - 5 - 6 647 82
          FQ225380.1 - 5 - 6 560 82
          FQ226971.1 - 5 - 6 606 82
          FQ227186.1 - 5 - 6 606 82
          FQ232999.1 - 2 - 3 165 82
          X13933.1 - 5 - 6 534 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Phosphodiesterase Inventoried Rat Phosphodiesterase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calm1

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          response to amphetamine
          regulation of heart rate
          detection of calcium ion
          activation of adenylate cyclase activity
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          calcium-mediated signaling
          substantia nigra development
          positive regulation of cyclic nucleotide metabolic process
          regulation of cytokinesis
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of protein dephosphorylation
          positive regulation of DNA binding
          positive regulation of nitric-oxide synthase activity
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          response to calcium ion
          regulation of cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          positive regulation of ryanodine-sensitive calcium-release channel activity
          regulation of store-operated calcium channel activity
          regulation of high voltage-gated calcium channel activity
          calcium channel regulator activity
          calcium ion binding
          protein binding
          adenylate cyclase binding
          protein kinase binding
          protein domain specific binding
          enzyme regulator activity
          nitric-oxide synthase regulator activity
          titin binding
          type 3 metabotropic glutamate receptor binding
          thioesterase binding
          N-terminal myristoylation domain binding
          phospholipase binding
          phosphatidylinositol 3-kinase binding
          ion channel binding
          protein N-terminus binding
          calcium-dependent protein binding
          nitric-oxide synthase binding
          protein phosphatase activator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P191784 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P015022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42037 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015022 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P015022 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P191784 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P015022 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191784 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P374988 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700146904 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700148336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78317 - -
          Agilent Rat cDNA Microarray (G4105A) 700148336 - -
          Agilent Rat cDNA Microarray (G4105A) 700146904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702198 - -
          Affymetrix Rat Gene 1.0 ST Array 10702197 - -
          Affymetrix Rat Gene 1.0 ST Array 10886382 - -
          Affymetrix Rat Gene 1.0 ST Array 10886379 - -
          Affymetrix Rat Gene 1.0 ST Array 10886381 - -
          Affymetrix Rat Gene 1.0 ST Array 10886383 - -
          Affymetrix Rat Gene 1.0 ST Array 10886380 - -
          Affymetrix Rat Gene 1.1 ST Array 10702196 - -
          Affymetrix Rat Gene 1.1 ST Array 10886378 - -
          Rat Genome 230 2.0 Array 1369936_PM_at - -
          Rat Genome 230 2.0 Array 1369936_at - -
          Rat Genome 230 2.0 Array 1387772_at - -
          Rat Genome 230 2.0 Array 1369937_PM_at - -
          Rat Genome 230 2.0 Array 1387772_PM_at - -
          Rat Genome 230 2.0 Array 1369937_at - -
          Affymetrix HT Rat Focus Array 1387772_at - -
          Affymetrix HT Rat Focus Array 1369936_at - -
          Affymetrix HT Rat Focus Array 1369937_PM_at - -
          Affymetrix HT Rat Focus Array 1369937_at - -
          Affymetrix HT Rat Focus Array 1387772_PM_at - -
          Affymetrix HT Rat Focus Array 1369936_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600009333.rn.1 TC0600001346.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600009334.rn.1 TC0600001346.rn.1 RTA TC
          Rat Expression Set 230 1369936_at - -
          Rat Expression Set 230 1369937_PM_at - -
          Rat Expression Set 230 1369937_at - -
          Rat Expression Set 230 1387772_PM_at - -
          Rat Expression Set 230 1387772_at - -
          Rat Expression Set 230 1369936_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10886378 - -
          Affymetrix Rat Gene 1.0 ST Array 10702196 - -
          Rat Genome U34 Set X13933_s_at - -
          Rat Genome U34 Set E02315cds_f_at - -
          Rat Genome U34 Set rc_AA849525_s_at - -
          Rat Genome U34 Set rc_AA849525_i_at - -
          Rat Genome U34 Set rc_AI171814_at - -
          Rat Genome U34 Set rc_AI180372_f_at - -
          Rat Genome U34 Set rc_AI102076_at - -
          Rat Genome U34 Set rc_AI229037_at - -
          Rat Genome U34 Set E02315cds_f_at - -
          Rat Genome U34 Set X13933_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline