Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821436_g1
          See other RPL21 GE Assays ›
          Gene Symbol:
          Rpl21
          Gene Name:
          ribosomal protein L21
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 9996919 - 9999998 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821436_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053330.1 NP_445782.1 6 494 82
          GenBank mRNA BC058459.1 - 6 498 6918772 82
          BC086441.1 - 6 487 7303531 82
          FQ217086.1 - 6 493 82
          FQ217504.1 - 6 492 82
          FQ222392.1 - 6 493 82
          FQ223144.1 - 6 492 82
          FQ224800.1 - 6 493 82
          FQ228557.1 - 6 493 82
          M27905.1 - 6 494 82
          X15216.1 - 6 494 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl21

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071317 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101917 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031573 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031573 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101917 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010514 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P495397 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487819 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255509 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P358632 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P473767 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700179121 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700486504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8571 - -
          Agilent Rat cDNA Microarray (G4105A) 700486504 - -
          Agilent Rat cDNA Microarray (G4105A) 700179121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884905 - -
          Affymetrix Rat Gene 1.0 ST Array 10923523 - -
          Affymetrix Rat Gene 1.0 ST Array 10792215 - -
          Affymetrix Rat Gene 1.0 ST Array 10879320 - -
          Affymetrix Rat Gene 1.0 ST Array 10917691 - -
          Affymetrix Rat Gene 1.0 ST Array 10928429 - -
          Affymetrix Rat Gene 1.0 ST Array 10833928 - -
          Affymetrix Rat Gene 1.0 ST Array 10847923 - -
          Affymetrix Rat Gene 1.0 ST Array 10880912 - -
          Affymetrix Rat Gene 1.0 ST Array 10812771 - -
          Affymetrix Rat Gene 1.0 ST Array 10870037 - -
          Affymetrix Rat Gene 1.0 ST Array 10910618 - -
          Affymetrix Rat Gene 1.0 ST Array 10723698 - -
          Affymetrix Rat Gene 1.0 ST Array 10760093 - -
          Affymetrix Rat Gene 1.0 ST Array 10804561 - -
          Affymetrix Rat Gene 1.0 ST Array 10785179 - -
          Affymetrix Rat Gene 1.0 ST Array 10859605 - -
          Affymetrix Rat Gene 1.0 ST Array 10821649 - -
          Affymetrix Rat Gene 1.0 ST Array 10730390 - -
          Affymetrix Rat Gene 1.0 ST Array 10788568 - -
          Affymetrix Rat Gene 1.0 ST Array 10793391 - -
          Affymetrix Rat Gene 1.0 ST Array 10776436 - -
          Affymetrix Rat Gene 1.0 ST Array 10885698 - -
          Affymetrix Rat Gene 1.0 ST Array 10856231 - -
          Affymetrix Rat Gene 1.0 ST Array 10937565 - -
          Affymetrix Rat Gene 1.0 ST Array 10799262 - -
          Affymetrix Rat Gene 1.0 ST Array 10799506 - -
          Affymetrix Rat Gene 1.0 ST Array 10766562 - -
          Affymetrix Rat Gene 1.0 ST Array 10798695 - -
          Affymetrix Rat Gene 1.0 ST Array 10911444 - -
          Affymetrix Rat Gene 1.0 ST Array 10870038 - -
          Affymetrix Rat Gene 1.0 ST Array 10745849 - -
          Affymetrix Rat Gene 1.0 ST Array 10928430 - -
          Affymetrix Rat Gene 1.0 ST Array 10788509 - -
          Affymetrix Rat Gene 1.0 ST Array 10712579 - -
          Affymetrix Rat Gene 1.0 ST Array 10855556 - -
          Affymetrix Rat Gene 1.0 ST Array 10838080 - -
          Affymetrix Rat Gene 1.0 ST Array 10760091 - -
          Affymetrix Rat Gene 1.0 ST Array 10801961 - -
          Affymetrix Rat Gene 1.0 ST Array 10760090 - -
          Affymetrix Rat Gene 1.0 ST Array 10870137 - -
          Affymetrix Rat Gene 1.0 ST Array 10760089 - -
          Affymetrix Rat Gene 1.0 ST Array 10821164 - -
          Affymetrix Rat Gene 1.0 ST Array 10760092 - -
          Affymetrix Rat Gene 1.0 ST Array 10838116 - -
          Affymetrix Rat Gene 1.0 ST Array 10817864 - -
          Affymetrix Rat Gene 1.0 ST Array 10928428 - -
          Affymetrix Rat Gene 1.0 ST Array 10826708 - -
          Affymetrix Rat Gene 1.0 ST Array 10826825 - -
          Affymetrix Rat Gene 1.0 ST Array 10762008 - -
          Affymetrix Rat Gene 1.0 ST Array 10846140 - -
          Affymetrix Rat Gene 1.0 ST Array 10782163 - -
          Affymetrix Rat Gene 1.0 ST Array 10908860 - -
          Affymetrix Rat Gene 1.0 ST Array 10767009 - -
          Affymetrix Rat Gene 1.0 ST Array 10861306 - -
          Affymetrix Rat Gene 1.0 ST Array 10878431 - -
          Affymetrix Rat Gene 1.0 ST Array 10782064 - -
          Affymetrix Rat Gene 1.0 ST Array 10932268 - -
          Affymetrix Rat Gene 1.0 ST Array 10930408 - -
          Affymetrix Rat Gene 1.1 ST Array 10930407 - -
          Affymetrix Rat Gene 1.1 ST Array 10712578 - -
          Affymetrix Rat Gene 1.1 ST Array 10923522 - -
          Affymetrix Rat Gene 1.1 ST Array 10766561 - -
          Affymetrix Rat Gene 1.1 ST Array 10878430 - -
          Affymetrix Rat Gene 1.1 ST Array 10838079 - -
          Affymetrix Rat Gene 1.1 ST Array 10847922 - -
          Affymetrix Rat Gene 1.1 ST Array 10870136 - -
          Affymetrix Rat Gene 1.1 ST Array 10782063 - -
          Affymetrix Rat Gene 1.1 ST Array 10937564 - -
          Affymetrix Rat Gene 1.1 ST Array 10838115 - -
          Affymetrix Rat Gene 1.1 ST Array 10932267 - -
          Affymetrix Rat Gene 1.1 ST Array 10782162 - -
          Affymetrix Rat Gene 1.1 ST Array 10879319 - -
          Affymetrix Rat Gene 1.1 ST Array 10767008 - -
          Affymetrix Rat Gene 1.1 ST Array 10760088 - -
          Affymetrix Rat Gene 1.1 ST Array 10884904 - -
          Affymetrix Rat Gene 1.1 ST Array 10793390 - -
          Affymetrix Rat Gene 1.1 ST Array 10928427 - -
          Affymetrix Rat Gene 1.1 ST Array 10911443 - -
          Affymetrix Rat Gene 1.1 ST Array 10788566 - -
          Affymetrix Rat Gene 1.1 ST Array 10855555 - -
          Affymetrix Rat Gene 1.1 ST Array 10826824 - -
          Affymetrix Rat Gene 1.1 ST Array 10833927 - -
          Affymetrix Rat Gene 1.1 ST Array 10799261 - -
          Affymetrix Rat Gene 1.1 ST Array 10776435 - -
          Affymetrix Rat Gene 1.1 ST Array 10861305 - -
          Affymetrix Rat Gene 1.1 ST Array 10745848 - -
          Affymetrix Rat Gene 1.1 ST Array 10846139 - -
          Affymetrix Rat Gene 1.1 ST Array 10799505 - -
          Affymetrix Rat Gene 1.1 ST Array 10730389 - -
          Affymetrix Rat Gene 1.1 ST Array 10859604 - -
          Affymetrix Rat Gene 1.1 ST Array 10908859 - -
          Affymetrix Rat Gene 1.1 ST Array 10792214 - -
          Affymetrix Rat Gene 1.1 ST Array 10723697 - -
          Affymetrix Rat Gene 1.1 ST Array 10885697 - -
          Affymetrix Rat Gene 1.1 ST Array 10821648 - -
          Affymetrix Rat Gene 1.1 ST Array 10817863 - -
          Affymetrix Rat Gene 1.1 ST Array 10798694 - -
          Affymetrix Rat Gene 1.1 ST Array 10788508 - -
          Affymetrix Rat Gene 1.1 ST Array 10821163 - -
          Affymetrix Rat Gene 1.1 ST Array 10826707 - -
          Affymetrix Rat Gene 1.1 ST Array 10785178 - -
          Affymetrix Rat Gene 1.1 ST Array 10917690 - -
          Affymetrix Rat Gene 1.1 ST Array 10880911 - -
          Affymetrix Rat Gene 1.1 ST Array 10812770 - -
          Affymetrix Rat Gene 1.1 ST Array 10856230 - -
          Affymetrix Rat Gene 1.1 ST Array 10870036 - -
          Rat Genome 230 2.0 Array 1398764_PM_at - -
          Rat Genome 230 2.0 Array 1398764_at - -
          Affymetrix HT Rat Focus Array 1398764_PM_at - -
          Affymetrix HT Rat Focus Array 1398764_at - -
          Affymetrix Rat Clariom S Assay PSR1200008919.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008923.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008926.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008918.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008922.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008924.rn.1 TC1200001057.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200008925.rn.1 TC1200001057.rn.1 RTA TC
          Rat Expression Set 230 1398764_at - -
          Rat Expression Set 230 1398764_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859604 - -
          Affymetrix Rat Gene 1.0 ST Array 10776435 - -
          Affymetrix Rat Gene 1.0 ST Array 10932267 - -
          Affymetrix Rat Gene 1.0 ST Array 10745848 - -
          Affymetrix Rat Gene 1.0 ST Array 10730389 - -
          Affymetrix Rat Gene 1.0 ST Array 10917690 - -
          Affymetrix Rat Gene 1.0 ST Array 10821163 - -
          Affymetrix Rat Gene 1.0 ST Array 10767008 - -
          Affymetrix Rat Gene 1.0 ST Array 10826824 - -
          Affymetrix Rat Gene 1.0 ST Array 10838115 - -
          Affymetrix Rat Gene 1.0 ST Array 10908859 - -
          Affymetrix Rat Gene 1.0 ST Array 10812770 - -
          Affymetrix Rat Gene 1.0 ST Array 10821648 - -
          Affymetrix Rat Gene 1.0 ST Array 10928427 - -
          Affymetrix Rat Gene 1.0 ST Array 10792214 - -
          Affymetrix Rat Gene 1.0 ST Array 10712578 - -
          Affymetrix Rat Gene 1.0 ST Array 10846139 - -
          Affymetrix Rat Gene 1.0 ST Array 10826707 - -
          Affymetrix Rat Gene 1.0 ST Array 10799505 - -
          Affymetrix Rat Gene 1.0 ST Array 10838079 - -
          Affymetrix Rat Gene 1.0 ST Array 10760088 - -
          Rat Genome U34 Set M27905_at - -
          Rat Genome U34 Set X15216cds_s_at - -
          Rat Genome U34 Set rc_AA849648_g_at - -
          Rat Genome U34 Set rc_AA849648_at - -
          Rat Genome U34 Set rc_AI070653_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349782 - -
          RatRef-12 Expression BeadChip ILMN_48068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline