Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821451_g1
          See other RAN GE Assays ›
          Gene Symbol:
          Ran
          Gene Name:
          RAN, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 31319556 - 31323758 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821451_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ran
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053439.1 NP_445891.1 4 - 5 325 91
          XM_006249285.2 XP_006249347.1 4 - 5 312 91
          GenBank mRNA AF306457.1 - 4 - 5 325 91
          BC059123.1 - 4 - 5 340 6920235 91
          FQ220326.1 - 4 - 5 308 91
          FQ220799.1 - 4 - 5 353 91
          FQ229342.1 - 4 - 5 277 91
          FQ229588.1 - 4 - 5 331 91
          FQ229761.1 - 4 - 5 316 91
          FQ231051.1 - 3 - 4 320 91
          FQ233248.1 - 4 - 5 279 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ran

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit export from nucleus
          ribosomal small subunit export from nucleus
          microtubule cytoskeleton organization
          protein import into nucleus
          protein export from nucleus
          nucleocytoplasmic transport
          mitotic nuclear division
          small GTPase mediated signal transduction
          spermatid development
          metabolic process
          actin cytoskeleton organization
          positive regulation of protein binding
          cellular protein localization
          cellular protein complex localization
          cell division
          GTPase activity
          GTP binding
          GDP binding
          protein domain specific binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859805 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12806 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12806 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12806 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761467 - -
          Affymetrix Rat Gene 1.0 ST Array 10703906 - -
          Affymetrix Rat Gene 1.0 ST Array 10761465 - -
          Affymetrix Rat Gene 1.0 ST Array 10761464 - -
          Affymetrix Rat Gene 1.0 ST Array 10761469 - -
          Affymetrix Rat Gene 1.0 ST Array 10761468 - -
          Affymetrix Rat Gene 1.0 ST Array 10761466 - -
          Affymetrix Rat Gene 1.1 ST Array 10703905 - -
          Affymetrix Rat Gene 1.1 ST Array 10761463 - -
          Rat Genome 230 2.0 Array 1367590_at - -
          Rat Genome 230 2.0 Array 1367590_PM_at - -
          Affymetrix HT Rat Focus Array 1367590_at - -
          Affymetrix HT Rat Focus Array 1367590_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200012384.rn.1 TC1200001464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012383.rn.1 TC1200001464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012379.rn.1 TC1200001464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012382.rn.1 TC1200001464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200012385.rn.1 TC1200001464.rn.1 RTA TC
          Rat Expression Set 230 1367590_at - -
          Rat Expression Set 230 1367590_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761463 - -
          Rat Genome U34 Set rc_AI175528_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22219374 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040754 - -
          RatRef-12 Expression BeadChip ILMN_62438 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline