Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821462_g1
          See other TMED10 GE Assays ›
          Gene Symbol:
          Tmed10
          Gene Name:
          transmembrane emp24-like trafficking protein 10 (yeast)
          Gene Aliases:
          Tmp21
          Chromosome Location:
          Chr.6: 109169739 - 109205004 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821462_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmed10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053467.1 NP_445919.1 2 - 3 342 111
          GenBank mRNA AJ004912.1 - 2 - 3 342 111
          X97443.1 - 2 - 3 294 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmed10

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          intracellular protein transport
          retrograde vesicle-mediated transport, Golgi to ER
          vesicle targeting
          Golgi organization
          COPI-coated vesicle budding
          response to alkaloid
          regulated exocytosis
          vesicle targeting, to, from or within Golgi
          protein oligomerization
          regulation of beta-amyloid formation
          protein binding
          syntaxin binding
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P671502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62057 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P671502 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P175391 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700693268 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47983 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50509 - -
          Agilent Rat cDNA Microarray (G4105A) 700693268 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891296 - -
          Affymetrix Rat Gene 1.0 ST Array 10891295 - -
          Affymetrix Rat Gene 1.0 ST Array 10886030 - -
          Affymetrix Rat Gene 1.0 ST Array 10891294 - -
          Affymetrix Rat Gene 1.1 ST Array 10886029 - -
          Affymetrix Rat Gene 1.1 ST Array 10891293 - -
          Rat Genome 230 2.0 Array 1398796_at - -
          Rat Genome 230 2.0 Array 1398796_PM_at - -
          Affymetrix HT Rat Focus Array 1398796_PM_at - -
          Affymetrix HT Rat Focus Array 1398796_at - -
          Affymetrix Rat Clariom S Assay PSR0600020419.rn.1 TC0600002870.rn.1 RTA TC
          Rat Expression Set 230 1398796_PM_at - -
          Rat Expression Set 230 1398796_at - -
          Rat Genome U34 Set X97443_s_at - -
          Rat Genome U34 Set AJ004912_s_at - -
          Rat Genome U34 Set rc_AA997494_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21466790 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352091 - -
          RatRef-12 Expression BeadChip ILMN_53887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline