Search
Search
| Gene Symbol: Kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270802.1 | NP_001257731.1 | 8 - 9 | 1297 | 62 | |
| NM_053483.2 | NP_445935.2 | 8 - 9 | 1272 | 62 | ||
| GenBank mRNA | AJ130946.1 | - | 7 - 8 | 1176 | 62 | |
| AY779026.1 | - | 7 - 8 | 1166 | 62 | ||
| BC062026.1 | - | 8 - 9 | 1272 | 5621957 | 62 | |
| BC089787.1 | - | 8 - 9 | 1223 | 7326072 | 62 | |
Set Membership: |
|