Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821482_g1
          See other ATP6V0E1 GE Assays ›
          Gene Symbol:
          Atp6v0e1
          Gene Name:
          ATPase, H+ transporting, lysosomal, V0 subunit e1
          Gene Aliases:
          Atp6k, Atp6v0e, Dsr-1A, dsr-1
          Chromosome Location:
          Chr.10: 16769857 - 16792930 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821482_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp6v0e1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053578.3 NP_446030.3 3 - 4 380 74
          GenBank mRNA AB037248.1 - 3 - 4 380 74
          AB051300.1 - 2 - 3 1522 74
          FQ226644.1 - 3 - 4 343 74
          FQ226841.1 - 3 - 4 350 74
          FQ228396.1 - 3 - 4 337 74
          FQ233078.1 - 3 - 4 339 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp6v0e1

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          ATP hydrolysis coupled proton transport
          proton transport
          cell growth
          response to amino acid
          transmembrane transport
          molecular_function
          proton-transporting ATPase activity, rotational mechanism

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P191467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191467 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P191467 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P191467 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P191467 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40426 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741843 - -
          Affymetrix Rat Gene 1.0 ST Array 10741842 - -
          Affymetrix Rat Gene 1.0 ST Array 10741845 - -
          Affymetrix Rat Gene 1.0 ST Array 10741846 - -
          Affymetrix Rat Gene 1.0 ST Array 10741844 - -
          Affymetrix Rat Gene 1.1 ST Array 10741841 - -
          Rat Genome 230 2.0 Array 1367724_a_at - -
          Rat Genome 230 2.0 Array 1367724_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367724_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367724_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000022604.rn.1 TC1000002487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000022606.rn.1 TC1000002487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000022607.rn.1 TC1000002487.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000022605.rn.1 TC1000002487.rn.1 RTA TC
          Rat Expression Set 230 1367724_a_at - -
          Rat Expression Set 230 1367724_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741841 - -
          Rat Genome U34 Set rc_AI013039_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20908296 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354260 - -
          RatRef-12 Expression BeadChip ILMN_50759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline