Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821506_g1
          See other KLF4 GE Assays ›
          Gene Symbol:
          Klf4
          Gene Name:
          Kruppel-like factor 4 (gut)
          Gene Aliases:
          GKLF
          Chromosome Location:
          Chr.5: 72283311 - 72287669 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821506_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053713.1 NP_446165.1 2 - 3 202 77
          GenBank mRNA AF390546.1 - 2 - 3 202 77
          BC085720.1 - 2 - 3 595 7190219 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002784.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf4

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of cell proliferation
          epidermal cell differentiation
          response to organic substance
          negative regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of muscle hyperplasia
          stem cell population maintenance
          cell differentiation
          post-embryonic camera-type eye development
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of cellular protein metabolic process
          response to retinoic acid
          negative regulation of heterotypic cell-cell adhesion
          post-embryonic hemopoiesis
          regulation of cell proliferation
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of phosphatidylinositol 3-kinase activity
          negative regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hemoglobin biosynthetic process
          negative regulation of smooth muscle cell proliferation
          epidermis morphogenesis
          positive regulation of protein metabolic process
          negative regulation of protein kinase B signaling
          positive regulation of telomerase activity
          canonical Wnt signaling pathway
          negative regulation of response to cytokine stimulus
          cellular response to hydrogen peroxide
          negative regulation of ERK1 and ERK2 cascade
          cellular response to retinoic acid
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          cellular response to cycloheximide
          cellular response to laminar fluid shear stress
          negative regulation of cell migration involved in sprouting angiogenesis
          cellular response to peptide
          negative regulation of chemokine (C-X-C motif) ligand 2 production
          core promoter proximal region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding transcription factor recruiting
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription cofactor binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          beta-catenin binding
          transcription factor binding
          phosphatidylinositol 3-kinase regulator activity
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12908 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12908 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12908 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72779 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876839 - -
          Affymetrix Rat Gene 1.0 ST Array 10876840 - -
          Affymetrix Rat Gene 1.0 ST Array 10873815 - -
          Affymetrix Rat Gene 1.0 ST Array 10876843 - -
          Affymetrix Rat Gene 1.0 ST Array 10876842 - -
          Affymetrix Rat Gene 1.1 ST Array 10876838 - -
          Affymetrix Rat Gene 1.1 ST Array 10873814 - -
          Rat Genome 230 2.0 Array 1387260_at - -
          Rat Genome 230 2.0 Array 1387260_PM_at - -
          Affymetrix HT Rat Focus Array 1387260_at - -
          Affymetrix HT Rat Focus Array 1387260_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500020563.rn.1 TC0500002784.rn.1 RTA TC
          Rat Expression Set 230 1387260_PM_at - -
          Rat Expression Set 230 1387260_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873814 - -
          Affymetrix Rat Gene 1.0 ST Array 10876838 - -
          Rat Genome U34 Set L26292_g_at - -
          Rat Genome U34 Set L26292_at - -
          Rat Genome U34 Set rc_AI179538_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20749852 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356110 - -
          RatRef-12 Expression BeadChip ILMN_55722 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline