Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821585_g1
          See other SLC25A5 GE Assays ›
          Gene Symbol:
          Slc25a5
          Gene Name:
          solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5
          Gene Aliases:
          Ant2
          Chromosome Location:
          Chr.X: 123404570 - 123407637 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821585_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057102.1 NP_476443.1 3 - 4 794 66
          GenBank mRNA BC059108.1 - 3 - 4 843 6922359 66
          D12771.1 - 3 - 4 794 66
          FQ213290.1 - 3 - 4 816 66
          FQ213855.1 - 3 - 4 834 66
          FQ214507.1 - 3 - 4 771 66
          FQ219677.1 - 3 - 4 816 66
          FQ220249.1 - 3 - 4 818 66
          FQ220272.1 - 3 - 4 819 66
          FQ220435.1 - 3 - 4 838 66
          FQ229325.1 - 3 - 4 819 66
          FQ229438.1 - 3 - 4 819 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc25a5

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          chromosome segregation
          positive regulation of cell proliferation
          transmembrane transport
          negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          transporter activity
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P175954 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P313659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P313659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P175954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P313659 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P313659 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P175954 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P175954 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P105366 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P280539 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700518703 - -
          Agilent Rat cDNA Microarray (G4105A) 700518703 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936317 - -
          Affymetrix Rat Gene 1.0 ST Array 10936315 - -
          Affymetrix Rat Gene 1.0 ST Array 10936316 - -
          Affymetrix Rat Gene 1.0 ST Array 10936314 - -
          Affymetrix Rat Gene 1.0 ST Array 10936318 - -
          Affymetrix Rat Gene 1.0 ST Array 10936313 - -
          Affymetrix Rat Gene 1.1 ST Array 10936312 - -
          Rat Genome 230 2.0 Array 1388163_PM_at - -
          Rat Genome 230 2.0 Array 1388163_at - -
          Affymetrix HT Rat Focus Array 1388163_PM_at - -
          Affymetrix HT Rat Focus Array 1388163_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006563.rn.1 TC0X00001059.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00006564.rn.1 TC0X00001059.rn.1 RTA TC
          Rat Expression Set 230 1388163_PM_at - -
          Rat Expression Set 230 1388163_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936312 - -
          Rat Genome U34 Set D12771_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22158328 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363856 - -
          RatRef-12 Expression BeadChip ILMN_48685 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline