Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821587_g1
          See other PRDX1 GE Assays ›
          Gene Symbol:
          Prdx1
          Gene Name:
          peroxiredoxin 1
          Gene Aliases:
          Hbp23
          Chromosome Location:
          Chr.5: 135536413 - 135551986 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821587_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057114.1 NP_476455.1 2 - 3 153 117
          GenBank mRNA BC058450.1 - 2 - 3 173 6920987 117
          BC088118.1 - 2 - 3 151 7324630 117
          D30035.1 - 2 - 3 153 117
          FQ219859.1 - 2 - 3 170 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx1

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          retina homeostasis
          response to oxidative stress
          cell proliferation
          removal of superoxide radicals
          regulation of stress-activated MAPK cascade
          erythrocyte homeostasis
          natural killer cell mediated cytotoxicity
          regulation of NF-kappaB import into nucleus
          hydrogen peroxide catabolic process
          oxidation-reduction process
          peroxidase activity
          thioredoxin peroxidase activity
          heme binding
          identical protein binding
          protein homodimerization activity
          poly(A) RNA binding
          peroxiredoxin activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P398167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170619 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398167 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101047 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818185 - -
          Affymetrix Rat Gene 1.0 ST Array 10871291 - -
          Affymetrix Rat Gene 1.0 ST Array 10871287 - -
          Affymetrix Rat Gene 1.0 ST Array 10871290 - -
          Affymetrix Rat Gene 1.0 ST Array 10871289 - -
          Affymetrix Rat Gene 1.0 ST Array 10871292 - -
          Affymetrix Rat Gene 1.0 ST Array 10871288 - -
          Affymetrix Rat Gene 1.1 ST Array 10818184 - -
          Affymetrix Rat Gene 1.1 ST Array 10871286 - -
          Rat Genome 230 2.0 Array 1367613_at - -
          Rat Genome 230 2.0 Array 1367613_PM_at - -
          Affymetrix HT Rat Focus Array 1367613_PM_at - -
          Affymetrix HT Rat Focus Array 1367613_at - -
          Affymetrix Rat Clariom S Assay PSR0500008801.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008803.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008805.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008804.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008802.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008806.rn.1 TC0500001270.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500008800.rn.1 TC0500001270.rn.1 RTA TC
          Rat Expression Set 230 1367613_at - -
          Rat Expression Set 230 1367613_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818184 - -
          Affymetrix Rat Gene 1.0 ST Array 10871286 - -
          Rat Genome U34 Set rc_AI010083_at - -
          Rat Genome U34 Set rc_AI177440_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21993834 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372596 - -
          RatRef-12 Expression BeadChip ILMN_68509 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline