Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821701_g1
          See other DNPH1 GE Assays ›
          Gene Symbol:
          Dnph1
          Gene Name:
          2'-deoxynucleoside 5'-phosphate N-hydrolase 1
          Gene Aliases:
          C6orf108, RGD620382, Rcl
          Chromosome Location:
          Chr.9: 16845764 - 16848514 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821701_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnph1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133525.1 NP_598209.1 1 - 2 184 73
          GenBank mRNA U82591.1 - 1 - 2 184 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnph1

          Panther Classification:

          Molecular Function -

          hydrolase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleoside metabolic process
          nucleotide metabolic process
          deoxyribonucleoside monophosphate catabolic process
          positive regulation of cell growth
          epithelial cell differentiation
          protein homodimerization activity
          nucleoside deoxyribosyltransferase activity
          deoxyribonucleoside 5'-monophosphate N-glycosidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P756652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90328 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P756652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P756652 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P756652 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P756652 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701912262 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700504992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35055 - -
          Agilent Rat cDNA Microarray (G4105A) 700504992 - -
          Agilent Rat cDNA Microarray (G4105A) 701912262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10926392 - -
          Affymetrix Rat Gene 1.0 ST Array 10926391 - -
          Affymetrix Rat Gene 1.0 ST Array 10926393 - -
          Affymetrix Rat Gene 1.0 ST Array 10926394 - -
          Affymetrix Rat Gene 1.1 ST Array 10926390 - -
          Rat Genome 230 2.0 Array 1368226_PM_at - -
          Rat Genome 230 2.0 Array 1368226_at - -
          Affymetrix HT Rat Focus Array 1368226_at - -
          Affymetrix HT Rat Focus Array 1368226_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900013090.rn.1 TC0900001569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900013091.rn.1 TC0900001569.rn.1 RTA TC
          Rat Expression Set 230 1368226_PM_at - -
          Rat Expression Set 230 1368226_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926390 - -
          Rat Genome U34 Set U82591_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20787779 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354636 - -
          RatRef-12 Expression BeadChip ILMN_51129 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline