Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821711_g1
          See other ATP5G2 GE Assays ›
          Gene Symbol:
          Atp5g2
          Gene Name:
          ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C2 (subunit 9)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 144264207 - 144272578 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821711_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp5g2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133556.2 NP_598240.1 3 - 4 161 98
          GenBank mRNA BC128726.1 - 3 - 4 161 7321526 98
          D13124.1 - 2 - 3 131 98
          FQ209535.1 - 170 98
          FQ235238.1 - 3 - 4 171 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp5g2

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          ATP synthesis coupled proton transport
          ATP hydrolysis coupled proton transport
          response to ethanol
          molecular_function
          lipid binding
          hydrogen ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038935 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P184527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15741 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P184527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038935 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P184527 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P184527 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038935 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P184527 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P841860 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700183041 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46628 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17384 - -
          Agilent Rat cDNA Microarray (G4105A) 700183041 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812588 - -
          Affymetrix Rat Gene 1.0 ST Array 10907635 - -
          Affymetrix Rat Gene 1.0 ST Array 10907636 - -
          Affymetrix Rat Gene 1.1 ST Array 10907634 - -
          Affymetrix Rat Gene 1.1 ST Array 10812587 - -
          Rat Genome 230 2.0 Array 1370207_PM_at - -
          Rat Genome 230 2.0 Array 1370207_at - -
          Affymetrix HT Rat Focus Array 1370207_PM_at - -
          Affymetrix HT Rat Focus Array 1370207_at - -
          Affymetrix Rat Clariom S Assay PSR0700032217.rn.1 TC0700003976.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032218.rn.1 TC0700003976.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032215.rn.1 TC0700003976.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032214.rn.1 TC0700003976.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032216.rn.1 TC0700003976.rn.1 RTA TC
          Rat Expression Set 230 1370207_PM_at - -
          Rat Expression Set 230 1370207_at - -
          Affymetrix Rat Gene 1.0 ST Array 10907634 - -
          Affymetrix Rat Gene 1.0 ST Array 10812587 - -
          Rat Genome U34 Set D13124_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21218577 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353161 - -
          RatRef-12 Expression BeadChip ILMN_56268 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline