Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821714_g1
          See other MPC1 GE Assays ›
          Gene Symbol:
          Mpc1
          Gene Name:
          mitochondrial pyruvate carrier 1
          Gene Aliases:
          Brp44l
          Chromosome Location:
          Chr.1: 53026608 - 53038229 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821714_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133561.1 NP_598245.1 4 - 5 372 73
          GenBank mRNA AF304429.1 - 4 - 5 372 73
          BC097287.1 - 4 - 5 388 7445460 73
          FQ219877.1 - 4 - 5 426 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mpc1

          Gene Ontology Categories:

          Function(s) Process(es)

          pyruvate metabolic process
          mitochondrial pyruvate transport
          pyruvate transmembrane transport
          pyruvate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085611 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085611 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P691927 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700861554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61229 - -
          Agilent Rat cDNA Microarray (G4105A) 700861554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718103 - -
          Affymetrix Rat Gene 1.0 ST Array 10718105 - -
          Affymetrix Rat Gene 1.0 ST Array 10718104 - -
          Affymetrix Rat Gene 1.0 ST Array 10718107 - -
          Affymetrix Rat Gene 1.1 ST Array 10718102 - -
          Rat Genome 230 2.0 Array 1386867_at - -
          Rat Genome 230 2.0 Array 1386867_PM_at - -
          Affymetrix HT Rat Focus Array 1386867_at - -
          Affymetrix HT Rat Focus Array 1386867_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100035721.rn.1 TC0100004504.rn.1 RTA TC
          Rat Expression Set 230 1386867_at - -
          Rat Expression Set 230 1386867_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718102 - -
          Rat Genome U34 Set U47315_s_at - -
          Rat Genome U34 Set rc_AI172414_at - -
          Rat Genome U34 Set U47315_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129417 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365165 - -
          RatRef-12 Expression BeadChip ILMN_65922 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline