Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821719_g1
          See other SUMO2 GE Assays ›
          Gene Symbol:
          Sumo2
          Gene Name:
          small ubiquitin-like modifier 2
          Gene Aliases:
          Smt3A, Smt3h2
          Chromosome Location:
          Chr.10: 104085401 - 104098216 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821719_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sumo2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133594.2 NP_598278.1 3 - 4 384 74
          XM_008768397.1 XP_008766619.1 4 - 5 524 74
          GenBank mRNA BC058446.1 - 347 6921523 74
          BC078746.1 - 3 - 4 318 7125132 74
          CF976332.1 - 388 74
          FQ221996.1 - 351 74
          FQ225379.1 - 326 74
          FQ225921.1 - 342 74
          FQ225989.1 - 342 74
          FQ235035.1 - 3 - 4 265 74
          L79949.1 - 3 - 4 331 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sumo2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein sumoylation
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          cellular protein localization
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          protein binding
          SUMO transferase activity
          ubiquitin protein ligase binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016048 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003664 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P016049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172347 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003664 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P016049 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003664 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016049 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P016048 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2720 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11478 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748971 - -
          Affymetrix Rat Gene 1.0 ST Array 10748972 - -
          Affymetrix Rat Gene 1.0 ST Array 10748970 - -
          Affymetrix Rat Gene 1.0 ST Array 10867410 - -
          Affymetrix Rat Gene 1.1 ST Array 10748969 - -
          Affymetrix Rat Gene 1.1 ST Array 10867409 - -
          Rat Genome 230 2.0 Array 1367452_at - -
          Rat Genome 230 2.0 Array 1367452_PM_at - -
          Affymetrix HT Rat Focus Array 1367452_at - -
          Affymetrix HT Rat Focus Array 1367452_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000037323.rn.1 TC1000004120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037324.rn.1 TC1000004120.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037325.rn.1 TC1000004120.rn.1 RTA TC
          Rat Expression Set 230 1367452_PM_at - -
          Rat Expression Set 230 1367452_at - -
          Rat Expression Set 230 1367452_PM_at - -
          Rat Expression Set 230 1367452_at - -
          Affymetrix Rat Gene 1.0 ST Array 10867409 - -
          Affymetrix Rat Gene 1.0 ST Array 10748969 - -
          Rat Genome U34 Set rc_AI010452_at - -
          Rat Genome U34 Set rc_AI233517_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22027385 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline