Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821765_g1
          See other VMP1 GE Assays ›
          Gene Symbol:
          Vmp1
          Gene Name:
          vacuole membrane protein 1
          Gene Aliases:
          Tmem49
          Chromosome Location:
          Chr.10: 73901988 - 74001895 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821765_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138839.2 NP_620194.1 4 - 5 415 102
          XM_006247078.2 XP_006247140.1 4 - 5 893 102
          XM_006247079.2 XP_006247141.1 5 - 6 437 102
          GenBank mRNA AF411216.1 - 4 - 5 361 102
          BC061721.1 - 4 - 5 415 5598561 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmp1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          autophagy
          endoplasmic reticulum organization
          Golgi organization
          embryo implantation
          regulation of autophagy
          single organismal cell-cell adhesion
          cell junction assembly
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P217541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145771 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P217541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P217541 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P217541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555389 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217541 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12160 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72553 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17219 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69663 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1729 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745935 - -
          Affymetrix Rat Gene 1.0 ST Array 10745937 - -
          Affymetrix Rat Gene 1.0 ST Array 10745938 - -
          Affymetrix Rat Gene 1.0 ST Array 10712103 - -
          Affymetrix Rat Gene 1.0 ST Array 10745936 - -
          Affymetrix Rat Gene 1.0 ST Array 10745939 - -
          Affymetrix Rat Gene 1.0 ST Array 10745934 - -
          Affymetrix Rat Gene 1.1 ST Array 10712101 - -
          Affymetrix Rat Gene 1.1 ST Array 10745933 - -
          Rat Genome 230 2.0 Array 1370807_at - -
          Rat Genome 230 2.0 Array 1383005_PM_at - -
          Rat Genome 230 2.0 Array 1383005_at - -
          Rat Genome 230 2.0 Array 1370807_PM_at - -
          Affymetrix HT Rat Focus Array 1370807_at - -
          Affymetrix HT Rat Focus Array 1370807_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000031109.rn.1 TC1000003470.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031108.rn.1 TC1000003470.rn.1 RTA TC
          Rat Expression Set 230 1370807_PM_at - -
          Rat Expression Set 230 1370807_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745933 - -
          Affymetrix Rat Gene 1.0 ST Array 10712101 - -
          Rat Genome U34 Set rc_AA859954_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21194818 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349318 - -
          RatRef-12 Expression BeadChip ILMN_57554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline