Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821777_g1
          See other PHF5A GE Assays ›
          Gene Symbol:
          Phf5a
          Gene Name:
          PHD finger protein 5A
          Gene Aliases:
          Ac2-246
          Chromosome Location:
          Chr.7: 123095286 - 123101851 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821777_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phf5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138888.1 NP_620243.1 3 - 4 243 96
          GenBank mRNA AF495522.1 - 3 - 4 243 96
          AY321339.1 - 259 96
          BC168144.1 - 3 - 4 261 8372591 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phf5a

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P571955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870183 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P571955 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P571955 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P571955 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P571960 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P571955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12281 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905653 - -
          Affymetrix Rat Gene 1.0 ST Array 10905652 - -
          Affymetrix Rat Gene 1.1 ST Array 10905651 - -
          Rat Genome 230 2.0 Array 1371463_PM_at - -
          Rat Genome 230 2.0 Array 1371463_at - -
          Affymetrix HT Rat Focus Array 1371463_at - -
          Affymetrix HT Rat Focus Array 1371463_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700028044.rn.1 TC0700003558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700028040.rn.1 TC0700003558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700028041.rn.1 TC0700003558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700028039.rn.1 TC0700003558.rn.1 RTA TC
          Rat Expression Set 230 1371463_PM_at - -
          Rat Expression Set 230 1371463_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905651 - -
          Rat Genome U34 Set rc_AA963008_s_at - -
          Rat Genome U34 Set rc_AI169723_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21442886 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367817 - -
          RatRef-12 Expression BeadChip ILMN_69265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline