Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821847_g1
          See other AURKA GE Assays ›
          Gene Symbol:
          Aurka
          Gene Name:
          aurora kinase A
          Gene Aliases:
          ARK-1, Stk6
          Chromosome Location:
          Chr.3: 170361529 - 170380277 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821847_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aurka
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153296.2 NP_695208.2 2 - 3 311 91
          XM_006235675.2 XP_006235737.1 3 - 4 390 91
          XM_006235676.2 XP_006235738.1 3 - 4 407 91
          GenBank mRNA AF537333.1 - 2 - 3 299 91
          BC104676.1 - 2 - 3 311 7453115 91
          FQ227101.1 - 3 - 4 397 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aurka

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic spindle organization
          microtubule cytoskeleton organization
          mitotic cell cycle
          protein phosphorylation
          spindle organization
          mitotic spindle organization
          spindle assembly involved in female meiosis I
          mitotic nuclear division
          mitotic centrosome separation
          meiotic nuclear division
          anterior/posterior axis specification
          cell projection organization
          regulation of protein stability
          negative regulation of protein binding
          response to estradiol
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cytokinesis
          histone-serine phosphorylation
          negative regulation of apoptotic process
          centrosome organization
          cell division
          centrosome localization
          protein localization to centrosome
          positive regulation of oocyte maturation
          neuron projection extension
          protein kinase activity
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          ATP binding
          protein kinase binding
          ubiquitin protein ligase binding
          histone serine kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P155186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63227 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P155186 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P155186 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11212 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852094 - -
          Affymetrix Rat Gene 1.0 ST Array 10852098 - -
          Affymetrix Rat Gene 1.0 ST Array 10852095 - -
          Affymetrix Rat Gene 1.0 ST Array 10852097 - -
          Affymetrix Rat Gene 1.0 ST Array 10852099 - -
          Affymetrix Rat Gene 1.0 ST Array 10852092 - -
          Affymetrix Rat Gene 1.0 ST Array 10852096 - -
          Affymetrix Rat Gene 1.0 ST Array 10852093 - -
          Affymetrix Rat Gene 1.1 ST Array 10852091 - -
          Rat Genome 230 2.0 Array 1376039_PM_at - -
          Rat Genome 230 2.0 Array 1376039_at - -
          Affymetrix HT Rat Focus Array 1376039_at - -
          Affymetrix HT Rat Focus Array 1376039_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300037599.rn.1 TC0300004925.rn.1 RTA TC
          Rat Expression Set 230 1376039_PM_at - -
          Rat Expression Set 230 1376039_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852091 - -
          Rat Genome U34 Set rc_AI030470_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20948589 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363543 - -
          RatRef-12 Expression BeadChip ILMN_64271 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline