Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00821946_g1
          See other RPL13A GE Assays ›
          Gene Symbol:
          Rpl13a
          Gene Name:
          ribosomal protein L13A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 101120711 - 101123401 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00821946_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl13a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173340.2 NP_775462.2 4 - 5 271 66
          GenBank mRNA BC086382.1 - 4 - 5 271 7301486 66
          EU000472.1 - 4 - 5 271 66
          FQ219828.1 - 1 - 2 100 66
          FQ219967.1 - 1 - 2 100 66
          FQ220858.1 - 1 - 2 100 66
          FQ221547.1 - 1 - 2 100 66
          FQ222716.1 - 1 - 2 95 66
          FQ222814.1 - 1 - 2 100 66
          FQ223363.1 - 1 - 2 100 66
          FQ229112.1 - 1 - 2 100 66
          FQ229269.1 - 1 - 2 100 66
          FQ229407.1 - 1 - 2 100 66
          FQ229499.1 - 1 - 2 100 66
          FQ229513.1 - 1 - 2 100 66
          FQ229538.1 - 1 - 2 100 66
          FQ229596.1 - 1 - 2 100 66
          FQ229947.1 - 1 - 2 100 66
          FQ232992.1 - 1 - 2 100 66
          FQ233205.1 - 1 - 2 100 66
          X68282.1 - 4 - 5 272 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl13a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          negative regulation of translation
          response to lipopolysaccharide
          regulation of homeostatic process
          macrophage chemotaxis
          lung morphogenesis
          cellular response to interferon-gamma
          negative regulation of formation of translation preinitiation complex
          mRNA binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123799 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123799 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123799 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P834588 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700768558 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700528619 - -
          Agilent Rat cDNA Microarray (G4105A) 700768558 - -
          Agilent Rat cDNA Microarray (G4105A) 700528619 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10833587 - -
          Affymetrix Rat Gene 1.0 ST Array 10797936 - -
          Affymetrix Rat Gene 1.1 ST Array 10721684 - -
          Affymetrix Rat Gene 1.1 ST Array 10797935 - -
          Affymetrix Rat Gene 1.1 ST Array 10833586 - -
          Rat Genome 230 2.0 Array 1393994_PM_at - -
          Rat Genome 230 2.0 Array 1393994_at - -
          Affymetrix Rat Clariom S Assay PSR0100043685.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043689.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043697.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043692.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043705.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043703.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043694.rn.1 TC0100005378.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043688.rn.1 TC0100005378.rn.1 RTA TC
          Rat Expression Set 230 1393994_at - -
          Rat Expression Set 230 1393994_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10833586 - -
          Affymetrix Rat Gene 1.0 ST Array 10797935 - -
          Rat Genome U34 Set rc_AA891713_at - -
          Rat Genome U34 Set rc_AI012805_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20801584 - -
          ABI Rat Genome Survey Microarray 22273325 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363266 - -
          RatRef-12 Expression BeadChip ILMN_62010 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline