Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00822100_gH
          See other GPX2 GE Assays ›
          Gene Symbol:
          Gpx2
          Gene Name:
          glutathione peroxidase 2
          Gene Aliases:
          GPX-GI, GSHPx-2, GSHPx-GI
          Chromosome Location:
          Chr.6: 99839960 - 99843245 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00822100_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183403.2 NP_899653.2 1 - 2 340 69
          GenBank mRNA BG664050.1 - 1 - 2 370 69
          CX570255.1 - 1 - 2 39 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpx2

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          negative regulation of inflammatory response to antigenic stimulus
          response to oxidative stress
          response to symbiotic bacterium
          interaction with symbiont
          oxidation-reduction process
          glutathione peroxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059565 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059565 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059565 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059565 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P500056 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P411743 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10890730 - -
          Affymetrix Rat Gene 1.0 ST Array 10890731 - -
          Affymetrix Rat Gene 1.0 ST Array 10890733 - -
          Affymetrix Rat Gene 1.0 ST Array 10890732 - -
          Affymetrix Rat Gene 1.1 ST Array 10890729 - -
          Rat Genome 230 2.0 Array 1374070_at - -
          Rat Genome 230 2.0 Array 1374070_PM_at - -
          Affymetrix HT Rat Focus Array 1374070_at - -
          Affymetrix HT Rat Focus Array 1374070_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600019202.rn.1 TC0600002699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019201.rn.1 TC0600002699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019203.rn.1 TC0600002699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600019204.rn.1 TC0600002699.rn.1 RTA TC
          Rat Expression Set 230 1374070_PM_at - -
          Rat Expression Set 230 1374070_at - -
          Affymetrix Rat Gene 1.0 ST Array 10890729 - -
          Rat Genome U34 Set rc_AA800587_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421875 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368854 - -
          RatRef-12 Expression BeadChip ILMN_65301 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline