Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00824607_g1
          See other ACP1 GE Assays ›
          Gene Symbol:
          Acp1
          Gene Name:
          acid phosphatase 1, soluble
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 49836064 - 49851714 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00824607_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135562.1 NP_001129034.1 5 - 6 471 69
          NM_021262.2 NP_067085.1 5 - 6 438 69
          XM_006239965.2 XP_006240027.1 5 - 6 467 69
          XM_008764572.1 XP_008762794.1 5 - 6 471 69
          GenBank mRNA AF171072.1 - 5 - 6 401 69
          BC062229.1 - 5 - 6 438 6918194 69
          BC158763.1 - 5 - 6 471 7109165 69
          BC161990.1 - 5 - 6 417 9032684 69
          FQ213063.1 - 5 - 6 418 69
          FQ214806.1 - 4 - 5 306 69
          FQ215147.1 - 5 - 6 409 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acp1

          Panther Classification:

          Molecular Function -

          protein phosphatase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          dephosphorylation
          peptidyl-tyrosine dephosphorylation
          acid phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          phosphatase activity
          SH3 domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P668972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P668972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P668972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P668972 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P668972 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700857567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103269 - -
          Agilent Rat cDNA Microarray (G4105A) 700857567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10932237 - -
          Affymetrix Rat Gene 1.0 ST Array 10889519 - -
          Affymetrix Rat Gene 1.0 ST Array 10889514 - -
          Affymetrix Rat Gene 1.0 ST Array 10890339 - -
          Affymetrix Rat Gene 1.0 ST Array 10889518 - -
          Affymetrix Rat Gene 1.0 ST Array 10889521 - -
          Affymetrix Rat Gene 1.0 ST Array 10889520 - -
          Affymetrix Rat Gene 1.1 ST Array 10932236 - -
          Affymetrix Rat Gene 1.1 ST Array 10889513 - -
          Affymetrix Rat Gene 1.1 ST Array 10890338 - -
          Rat Genome 230 2.0 Array 1387840_at - -
          Rat Genome 230 2.0 Array 1398938_PM_at - -
          Rat Genome 230 2.0 Array 1387840_PM_at - -
          Rat Genome 230 2.0 Array 1398938_at - -
          Affymetrix HT Rat Focus Array 1398938_at - -
          Affymetrix HT Rat Focus Array 1387840_at - -
          Affymetrix HT Rat Focus Array 1387840_PM_at - -
          Affymetrix HT Rat Focus Array 1398938_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600016353.rn.1 TC0600002267.rn.1 RTA TC
          Rat Expression Set 230 1387840_PM_at - -
          Rat Expression Set 230 1387840_at - -
          Rat Expression Set 230 1398938_PM_at - -
          Rat Expression Set 230 1398938_at - -
          Affymetrix Rat Gene 1.0 ST Array 10889513 - -
          Affymetrix Rat Gene 1.0 ST Array 10932236 - -
          Affymetrix Rat Gene 1.0 ST Array 10890338 - -
          Rat Genome U34 Set rc_AI170780_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20779970 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361506 - -
          RatRef-12 Expression BeadChip ILMN_70348 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline