Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00824608_m1
          See other NXPH4 GE Assays ›
          Gene Symbol:
          Nxph4
          Gene Name:
          neurexophilin 4
          Gene Aliases:
          Nph4
          Chromosome Location:
          Chr.7: 70833662 - 70842405 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00824608_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxph4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021680.2 NP_067712.2 1 - 2 74 68
          GenBank mRNA AF042714.1 - 1 - 2 145 68
          BC081805.1 - 1 - 2 74 7108207 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002924.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nxph4

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P173451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91423 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139554 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P139554 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P139554 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903079 - -
          Affymetrix Rat Gene 1.0 ST Array 10903076 - -
          Affymetrix Rat Gene 1.0 ST Array 10903080 - -
          Affymetrix Rat Gene 1.0 ST Array 10903081 - -
          Affymetrix Rat Gene 1.0 ST Array 10903078 - -
          Affymetrix Rat Gene 1.0 ST Array 10903075 - -
          Affymetrix Rat Gene 1.0 ST Array 10903077 - -
          Affymetrix Rat Gene 1.1 ST Array 10903074 - -
          Rat Genome 230 2.0 Array 1387525_at - -
          Rat Genome 230 2.0 Array 1387525_PM_at - -
          Affymetrix HT Rat Focus Array 1387525_PM_at - -
          Affymetrix HT Rat Focus Array 1387525_at - -
          Affymetrix Rat Clariom S Assay PSR0700022629.rn.1 TC0700002924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022633.rn.1 TC0700002924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022630.rn.1 TC0700002924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022632.rn.1 TC0700002924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022634.rn.1 TC0700002924.rn.1 RTA TC
          Rat Expression Set 230 1387525_PM_at - -
          Rat Expression Set 230 1387525_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903074 - -
          Rat Genome U34 Set AF042714_at - -
          Rat Genome U34 Set rc_AA957158_at - -
          Rat Genome U34 Set AF042714_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21607468 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361576 - -
          RatRef-12 Expression BeadChip ILMN_50307 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline