Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00824644_g1
          See other ATF4 GE Assays ›
          Gene Symbol:
          Atf4
          Gene Name:
          activating transcription factor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121480723 - 121482781 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00824644_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024403.2 NP_077379.1 2 - 3 491 68
          GenBank mRNA AF252627.1 - 1 - 2 257 68
          BC158588.1 - 2 - 3 464 9098368 68
          FQ210065.1 - 2 - 3 491 68
          FQ213021.1 - 2 - 3 491 68
          FQ214151.1 - 2 - 3 490 68
          FQ214217.1 - 2 - 3 492 68
          FQ214457.1 - 2 - 3 490 68
          FQ216049.1 - 2 - 3 490 68
          FQ219043.1 - 2 - 3 491 68
          FQ221881.1 - 2 - 3 491 68
          FQ230257.1 - 1 - 2 119 68
          FQ232693.1 - 2 - 3 497 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004036.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf4

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          gluconeogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          gamma-aminobutyric acid signaling pathway
          positive regulation of vascular endothelial growth factor production
          circadian regulation of gene expression
          response to endoplasmic reticulum stress
          positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          cellular response to glucose starvation
          negative regulation of potassium ion transport
          positive regulation of neuron apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase I promoter
          positive regulation of transcription from RNA polymerase II promoter
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of oxidative stress-induced neuron death
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein C-terminus binding
          transcription factor binding
          leucine zipper domain binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P620762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P620762 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P620762 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P620762 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P620762 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897762 - -
          Affymetrix Rat Gene 1.0 ST Array 10897763 - -
          Affymetrix Rat Gene 1.0 ST Array 10897761 - -
          Affymetrix Rat Gene 1.1 ST Array 10897760 - -
          Rat Genome 230 2.0 Array 1367624_PM_at - -
          Rat Genome 230 2.0 Array 1367624_at - -
          Affymetrix HT Rat Focus Array 1367624_PM_at - -
          Affymetrix HT Rat Focus Array 1367624_at - -
          Affymetrix Rat Clariom S Assay PSR0700032962.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032963.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032964.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032965.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032968.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032967.rn.1 TC0700004036.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032969.rn.1 TC0700004036.rn.1 RTA TC
          Rat Expression Set 230 1367624_at - -
          Rat Expression Set 230 1367624_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897760 - -
          Rat Genome U34 Set rc_AI012335_at - -
          Rat Genome U34 Set rc_AI179381_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21827490 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375100 - -
          RatRef-12 Expression BeadChip ILMN_53353 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline