Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01136020_gH
          See other GGPS1 GE Assays ›
          Gene Symbol:
          Ggps1
          Gene Name:
          geranylgeranyl diphosphate synthase 1
          Gene Aliases:
          Crlf3, rGGPS1a, rGGPS1a1, rGGPS1a2, rGGPS1a3
          Chromosome Location:
          Chr.17: 53956185 - 53971039 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01136020_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggps1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007626.2 NP_001007627.1 3 - 4 387 79
          XM_008771698.1 XP_008769920.1 3 - 4 405 79
          XM_008771699.1 XP_008769921.1 2 - 3 312 79
          GenBank mRNA AB118238.1 - 3 - 4 373 79
          AB118239.1 - 3 - 4 373 79
          AB118240.1 - 3 - 4 373 79
          AB118241.1 - 3 - 4 373 79
          AB118242.1 - 3 - 4 373 79
          AB118243.1 - 3 - 4 373 79
          BC090327.1 - 3 - 4 387 7125199 79

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_008771699.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggps1

          Gene Ontology Categories:

          Function(s) Process(es)

          isoprenoid metabolic process
          geranyl diphosphate biosynthetic process
          geranylgeranyl diphosphate biosynthetic process
          farnesyl diphosphate biosynthetic process
          dimethylallyltranstransferase activity
          farnesyltranstransferase activity
          geranyltranstransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026399 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026399 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026399 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P119296 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795526 - -
          Affymetrix Rat Gene 1.0 ST Array 10795523 - -
          Affymetrix Rat Gene 1.0 ST Array 10795524 - -
          Affymetrix Rat Gene 1.0 ST Array 10795525 - -
          Affymetrix Rat Gene 1.0 ST Array 10795521 - -
          Affymetrix Rat Gene 1.0 ST Array 10795522 - -
          Affymetrix Rat Gene 1.0 ST Array 10795527 - -
          Affymetrix Rat Gene 1.1 ST Array 10795520 - -
          Rat Genome 230 2.0 Array 1377866_a_at - -
          Rat Genome 230 2.0 Array 1380586_at - -
          Rat Genome 230 2.0 Array 1386215_at - -
          Rat Genome 230 2.0 Array 1380586_PM_at - -
          Rat Genome 230 2.0 Array 1379556_PM_a_at - -
          Rat Genome 230 2.0 Array 1386215_PM_at - -
          Rat Genome 230 2.0 Array 1379556_a_at - -
          Rat Genome 230 2.0 Array 1377866_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386215_PM_at - -
          Affymetrix HT Rat Focus Array 1377866_a_at - -
          Affymetrix HT Rat Focus Array 1377866_PM_a_at - -
          Affymetrix HT Rat Focus Array 1379556_a_at - -
          Affymetrix HT Rat Focus Array 1386215_at - -
          Affymetrix HT Rat Focus Array 1379556_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1700005176.rn.1 TC1700000774.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005179.rn.1 TC1700000774.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005177.rn.1 TC1700000774.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005178.rn.1 TC1700000774.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700005180.rn.1 TC1700000774.rn.1 RTA TC
          Rat Expression Set 230 1379556_PM_a_at - -
          Rat Expression Set 230 1380586_at - -
          Rat Expression Set 230 1377866_PM_a_at - -
          Rat Expression Set 230 1386215_at - -
          Rat Expression Set 230 1380586_PM_at - -
          Rat Expression Set 230 1377866_a_at - -
          Rat Expression Set 230 1379556_a_at - -
          Rat Expression Set 230 1386215_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795520 - -
          Rat Genome U34 Set rc_AI175248_at - -
          Rat Genome U34 Set rc_AI105210_at - -
          Rat Genome U34 Set rc_AI232147_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21058126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349919 - -
          RatRef-12 Expression BeadChip ILMN_50115 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline