Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01139424_m1
          See other CXXC5 GE Assays ›
          Gene Symbol:
          Cxxc5
          Gene Name:
          CXXC finger protein 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 28653653 - 28656908 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01139424_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxxc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007628.2 NP_001007629.1 1 - 2 1040 90
          GenBank mRNA BC083622.1 - 1 - 2 1040 7134100 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000294.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxxc5

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          DNA binding
          signal transducer activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P634770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201930 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P634770 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P634770 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P634770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10935257 - -
          Affymetrix Rat Gene 1.0 ST Array 10935259 - -
          Affymetrix Rat Gene 1.0 ST Array 10800971 - -
          Affymetrix Rat Gene 1.0 ST Array 10935258 - -
          Affymetrix Rat Gene 1.0 ST Array 10800972 - -
          Affymetrix Rat Gene 1.0 ST Array 10800970 - -
          Affymetrix Rat Gene 1.0 ST Array 10935256 - -
          Affymetrix Rat Gene 1.1 ST Array 10935255 - -
          Affymetrix Rat Gene 1.1 ST Array 10800969 - -
          Rat Genome 230 2.0 Array 1375231_PM_a_at - -
          Rat Genome 230 2.0 Array 1371629_PM_at - -
          Rat Genome 230 2.0 Array 1371629_at - -
          Rat Genome 230 2.0 Array 1375231_a_at - -
          Affymetrix HT Rat Focus Array 1375231_a_at - -
          Affymetrix HT Rat Focus Array 1371629_at - -
          Affymetrix HT Rat Focus Array 1371629_PM_at - -
          Affymetrix HT Rat Focus Array 1375231_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1800002213.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002220.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002216.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002218.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002217.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002221.rn.1 TC1800000294.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800002219.rn.1 TC1800000294.rn.1 RTA TC
          Rat Expression Set 230 1371629_at - -
          Rat Expression Set 230 1375231_a_at - -
          Rat Expression Set 230 1375231_PM_a_at - -
          Rat Expression Set 230 1371629_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10935255 - -
          Affymetrix Rat Gene 1.0 ST Array 10800969 - -
          Rat Genome U34 Set rc_H33001_at - -
          Rat Genome U34 Set rc_AA851147_at - -
          Rat Genome U34 Set rc_AI105444_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21306433 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360192 - -
          RatRef-12 Expression BeadChip ILMN_50285 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline