Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01151962_m1
          See other ELAVL3 GE Assays ›
          Gene Symbol:
          Elavl3
          Gene Name:
          ELAV like neuron-specific RNA binding protein 3
          Gene Aliases:
          Huc
          Chromosome Location:
          Chr.8: 23029507 - 23063359 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01151962_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Elavl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172324.1 NP_758827.1 3 - 4 374 66
          XM_006242616.2 XP_006242678.1 3 - 4 718 66
          XM_008765920.1 XP_008764142.1 3 - 4 561 66
          GenBank mRNA AB097198.1 - 3 - 4 374 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002093.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Elavl3

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          AU-rich element binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147374 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125876 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461122 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915710 - -
          Affymetrix Rat Gene 1.0 ST Array 10915715 - -
          Affymetrix Rat Gene 1.0 ST Array 10915713 - -
          Affymetrix Rat Gene 1.0 ST Array 10915711 - -
          Affymetrix Rat Gene 1.0 ST Array 10915709 - -
          Affymetrix Rat Gene 1.0 ST Array 10915714 - -
          Affymetrix Rat Gene 1.0 ST Array 10915712 - -
          Affymetrix Rat Gene 1.1 ST Array 10915708 - -
          Rat Genome 230 2.0 Array 1382169_PM_x_at - -
          Rat Genome 230 2.0 Array 1382168_at - -
          Rat Genome 230 2.0 Array 1382168_PM_at - -
          Rat Genome 230 2.0 Array 1382169_x_at - -
          Affymetrix HT Rat Focus Array 1382168_PM_at - -
          Affymetrix HT Rat Focus Array 1382168_at - -
          Affymetrix HT Rat Focus Array 1382169_x_at - -
          Affymetrix HT Rat Focus Array 1382169_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR0800016995.rn.1 TC0800002093.rn.1 RTA TC
          Rat Expression Set 230 1382169_PM_x_at - -
          Rat Expression Set 230 1382169_x_at - -
          Rat Expression Set 230 1382168_PM_at - -
          Rat Expression Set 230 1382168_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915708 - -
          Rat Genome U34 Set rc_AI044893_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21122228 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369333 - -
          RatRef-12 Expression BeadChip ILMN_52672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline