Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01158001_m1
          See other RGD1306271 GE Assays ›
          Gene Symbol:
          RGD1306271
          Gene Name:
          similar to KIAA1549 protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 65834933 - 65962722 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01158001_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1306271
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006236338.2 XP_006236400.1 18 - 19 5516 57
          XM_006236341.2 XP_006236403.1 18 - 19 7162 57
          XM_008762846.1 XP_008761068.1 18 - 19 6733 57
          XM_008762847.1 XP_008761069.1 18 - 19 5479 57
          XM_231616.8 XP_231616.5 18 - 19 5519 57

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P225050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861961 - -
          Affymetrix Rat Gene 1.0 ST Array 10861959 - -
          Affymetrix Rat Gene 1.0 ST Array 10861949 - -
          Affymetrix Rat Gene 1.0 ST Array 10861947 - -
          Affymetrix Rat Gene 1.0 ST Array 10861966 - -
          Affymetrix Rat Gene 1.0 ST Array 10861952 - -
          Affymetrix Rat Gene 1.0 ST Array 10861962 - -
          Affymetrix Rat Gene 1.0 ST Array 10861950 - -
          Affymetrix Rat Gene 1.0 ST Array 10861964 - -
          Affymetrix Rat Gene 1.0 ST Array 10861954 - -
          Affymetrix Rat Gene 1.0 ST Array 10861960 - -
          Affymetrix Rat Gene 1.0 ST Array 10861951 - -
          Affymetrix Rat Gene 1.0 ST Array 10861958 - -
          Affymetrix Rat Gene 1.0 ST Array 10861953 - -
          Affymetrix Rat Gene 1.0 ST Array 10861955 - -
          Affymetrix Rat Gene 1.0 ST Array 10861957 - -
          Affymetrix Rat Gene 1.0 ST Array 10861948 - -
          Affymetrix Rat Gene 1.0 ST Array 10861965 - -
          Affymetrix Rat Gene 1.0 ST Array 10861956 - -
          Affymetrix Rat Gene 1.0 ST Array 10861963 - -
          Affymetrix Rat Gene 1.0 ST Array 10861946 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21597997 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374545 - -
          RatRef-12 Expression BeadChip ILMN_70110 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0