Search
Search
| Gene Symbol: Dgki | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198782.2 | NP_942077.2 | 7 - 8 | 1103 | 61 | |
| XM_006236333.2 | XP_006236395.1 | 7 - 8 | 925 | 61 | ||
| GenBank mRNA | AB058962.1 | - | 7 - 8 | 1103 | 61 | |
| AB058963.1 | - | 7 - 8 | 1103 | 61 | ||
| AB058964.1 | - | 7 - 8 | 1103 | 61 | ||
Set Membership: |
|