Search
Search
| Gene Symbol: Celf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135603.1 | NP_001129075.1 | 8 - 9 | 1009 | 69 | |
| XM_006240941.2 | XP_006241003.2 | 8 - 9 | 1003 | 69 | ||
| XM_006240942.2 | XP_006241004.2 | 8 - 9 | 976 | 69 | ||
| XM_008765104.1 | XP_008763326.1 | 7 - 8 | 812 | 69 | ||
| XM_008765105.1 | XP_008763327.1 | 8 - 9 | 1001 | 69 | ||
| XM_008765106.1 | XP_008763328.1 | 8 - 9 | 1003 | 69 | ||
Set Membership: |
|