Search
Search
| Gene Symbol: Zmym4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107982.1 | NP_001101452.1 | 3 - 4 | 949 | 123 | |
| XM_006238873.1 | XP_006238935.1 | 3 - 4 | 944 | 123 | ||
| XM_008764026.1 | XP_008762248.1 | 3 - 4 | 944 | 123 | ||
Set Membership: |
|