Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01188200_m1
          See other NEPN GE Assays ›
          Gene Symbol:
          Nepn
          Gene Name:
          nephrocan
          Gene Aliases:
          RGD1307755
          Chromosome Location:
          Chr.20: 33527932 - 33543860 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01188200_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nepn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107632.1 NP_001101102.1 2 - 3 1018 63
          XM_006256491.2 XP_006256553.1 2 - 3 1303 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000721.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s)

          negative regulation of transcription from RNA polymerase II promoter
          extracellular matrix organization
          negative regulation of transforming growth factor beta receptor signaling pathway

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P115826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144086 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P115826 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P115826 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P295350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830153 - -
          Affymetrix Rat Gene 1.0 ST Array 10830154 - -
          Affymetrix Rat Gene 1.0 ST Array 10830152 - -
          Affymetrix Rat Gene 1.1 ST Array 10830149 - -
          Rat Genome 230 2.0 Array 1385115_at - -
          Rat Genome 230 2.0 Array 1385115_PM_at - -
          Affymetrix HT Rat Focus Array 1385115_PM_at - -
          Affymetrix HT Rat Focus Array 1385115_at - -
          Affymetrix Rat Clariom S Assay PSR2000006227.rn.1 TC2000000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006228.rn.1 TC2000000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000006225.rn.1 TC2000000721.rn.1 RTA TC
          Rat Expression Set 230 1385115_PM_at - -
          Rat Expression Set 230 1385115_at - -
          Affymetrix Rat Gene 1.0 ST Array 10830149 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21991395 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline