Search
Search
| Gene Symbol: Hdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053447.1 | NP_445899.1 | 9 - 10 | 1181 | 86 | |
| GenBank mRNA | AF321130.1 | - | 1 - 2 | 39 | 86 | |
| BC161939.1 | - | 9 - 10 | 1156 | 86 | ||
Set Membership: |
|