Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01207840_m1
          See other FAM83A GE Assays ›
          Gene Symbol:
          Fam83a
          Gene Name:
          family with sequence similarity 83, member A
          Gene Aliases:
          RGD1565606
          Chromosome Location:
          Chr.7: 97853091 - 97880936 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01207840_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam83a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130576.1 NP_001124048.1 3 - 4 929 95
          XM_008765518.1 XP_008763740.1 4 - 5 3564 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001151.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308897 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155621 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100384 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061825 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061825 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10896663 - -
          Affymetrix Rat Gene 1.0 ST Array 10896665 - -
          Affymetrix Rat Gene 1.0 ST Array 10896664 - -
          Affymetrix Rat Gene 1.0 ST Array 10896662 - -
          Affymetrix Rat Gene 1.1 ST Array 10896661 - -
          Affymetrix Rat Clariom S Assay PSR0700008409.rn.1 TC0700001151.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10896661 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21785412 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350319 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline