Search
Search
| Gene Symbol: Myt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108615.1 | NP_001102085.1 | 5 - 6 | 2222 | 59 | |
| XM_006235782.1 | XP_006235844.1 | 11 - 12 | 2039 | 59 | ||
| XM_006235783.2 | XP_006235845.1 | 11 - 12 | 2273 | 59 | ||
| XM_006235784.1 | XP_006235846.1 | 11 - 12 | 2039 | 59 | ||
| XM_006235785.2 | XP_006235847.1 | 11 - 12 | 1990 | 59 | ||
| XM_006235786.2 | XP_006235848.1 | 11 - 12 | 2039 | 59 | ||
| XM_006235787.2 | XP_006235849.1 | 8 - 9 | 1860 | 59 | ||
| XM_008762522.1 | XP_008760744.1 | 11 - 12 | 2039 | 59 | ||
Set Membership: |
|