Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01218287_m1
          See other SYT11 GE Assays ›
          Gene Symbol:
          Syt11
          Gene Name:
          synaptotagmin XI
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 188112301 - 188138180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01218287_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syt11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031667.2 NP_113855.2 2 - 3 1060 83
          XM_006232748.1 XP_006232810.1 2 - 3 1052 83
          GenBank mRNA AF000423.1 - 2 - 3 1101 83
          AF375465.1 - 2 - 3 859 83
          BC094518.1 - 2 - 3 1060 7315033 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004047.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Syt11

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity
          calcium ion binding
          calcium-dependent phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P663085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137535 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P663085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P663085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P663085 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P455693 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702080105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55038 - -
          Agilent Rat cDNA Microarray (G4105A) 702080105 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824352 - -
          Affymetrix Rat Gene 1.0 ST Array 10824356 - -
          Affymetrix Rat Gene 1.0 ST Array 10824355 - -
          Affymetrix Rat Gene 1.0 ST Array 10824353 - -
          Affymetrix Rat Gene 1.0 ST Array 10824354 - -
          Affymetrix Rat Gene 1.0 ST Array 10824350 - -
          Affymetrix Rat Gene 1.0 ST Array 10824351 - -
          Affymetrix Rat Gene 1.1 ST Array 10824349 - -
          Rat Genome 230 2.0 Array 1369135_PM_at - -
          Rat Genome 230 2.0 Array 1369135_at - -
          Affymetrix HT Rat Focus Array 1369135_at - -
          Affymetrix HT Rat Focus Array 1369135_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200027417.rn.1 TC0200004047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027419.rn.1 TC0200004047.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027420.rn.1 TC0200004047.rn.1 RTA TC
          Rat Expression Set 230 1369135_PM_at - -
          Rat Expression Set 230 1369135_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824349 - -
          Rat Genome U34 Set AF000423_at - -
          Rat Genome U34 Set AF000423_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363764 - -
          RatRef-12 Expression BeadChip ILMN_55156 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline