Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01218292_m1
          See other RGD1565775 GE Assays ›
          Gene Symbol:
          RGD1565775
          Gene Name:
          similar to RIKEN cDNA 2810403A07
          Gene Aliases:
          Blom7
          Chromosome Location:
          Chr.2: 188051659 - 188080280 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01218292_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1565775
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_342277.6 XP_342278.5 12 - 13 1593 57
          GenBank mRNA BC092205.1 - 12 - 13 1546 57
          BC103646.1 - 12 - 13 1543 57

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Process(es)

          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052335 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052335 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052335 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052335 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P181050 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33838 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816599 - -
          Affymetrix Rat Gene 1.0 ST Array 10816598 - -
          Affymetrix Rat Gene 1.0 ST Array 10816600 - -
          Affymetrix Rat Gene 1.0 ST Array 10816603 - -
          Affymetrix Rat Gene 1.0 ST Array 10816593 - -
          Affymetrix Rat Gene 1.0 ST Array 10816592 - -
          Affymetrix Rat Gene 1.0 ST Array 10816601 - -
          Affymetrix Rat Gene 1.0 ST Array 10816596 - -
          Affymetrix Rat Gene 1.0 ST Array 10816597 - -
          Affymetrix Rat Gene 1.0 ST Array 10816595 - -
          Affymetrix Rat Gene 1.0 ST Array 10816594 - -
          Affymetrix Rat Gene 1.0 ST Array 10816604 - -
          Affymetrix Rat Gene 1.0 ST Array 10816602 - -
          Affymetrix Rat Gene 1.1 ST Array 10816590 - -
          Rat Genome 230 2.0 Array 1373474_PM_at - -
          Rat Genome 230 2.0 Array 1373474_at - -
          Affymetrix HT Rat Focus Array 1373474_PM_at - -
          Affymetrix HT Rat Focus Array 1373474_at - -
          Rat Expression Set 230 1373474_PM_at - -
          Rat Expression Set 230 1373474_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816590 - -
          Rat Genome U34 Set rc_AA957205_at - -
          Rat Genome U34 Set rc_AI229179_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354105 - -
          RatRef-12 Expression BeadChip ILMN_63363 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline