Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01218688_m1
          See other N4BP2L2 GE Assays ›
          Gene Symbol:
          N4bp2l2
          Gene Name:
          NEDD4 binding protein 2-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 627128 - 650387 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01218688_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: N4bp2l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005533.1 NP_001005533.1 2 - 3 1327 61
          XM_006248735.2 XP_006248797.1 2 - 3 1313 61
          XM_006248736.2 XP_006248798.1 2 - 3 1338 61
          GenBank mRNA BC081998.1 - 2 - 3 1327 7131024 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000904.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene N4bp2l2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          blastocyst development
          transcription from RNA polymerase II promoter
          positive regulation of hematopoietic stem cell proliferation
          negative regulation of hematopoietic stem cell differentiation
          RNA polymerase II transcription corepressor activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P990142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P990142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P990142 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P990142 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P990142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66590 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21564 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756315 - -
          Affymetrix Rat Gene 1.0 ST Array 10756310 - -
          Affymetrix Rat Gene 1.0 ST Array 10756309 - -
          Affymetrix Rat Gene 1.0 ST Array 10756313 - -
          Affymetrix Rat Gene 1.0 ST Array 10756316 - -
          Affymetrix Rat Gene 1.0 ST Array 10756311 - -
          Affymetrix Rat Gene 1.0 ST Array 10756314 - -
          Affymetrix Rat Gene 1.1 ST Array 10756308 - -
          Rat Genome 230 2.0 Array 1390514_at - -
          Rat Genome 230 2.0 Array 1390514_PM_at - -
          Affymetrix HT Rat Focus Array 1390514_PM_at - -
          Affymetrix HT Rat Focus Array 1390514_at - -
          Affymetrix Rat Clariom S Assay PSR1200007914.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007918.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007916.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007919.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007912.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007913.rn.1 TC1200000904.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200007915.rn.1 TC1200000904.rn.1 RTA TC
          Rat Expression Set 230 1390514_at - -
          Rat Expression Set 230 1390514_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756308 - -
          Rat Genome U34 Set rc_AI058990_at - -
          Rat Genome U34 Set rc_AI044628_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362176 - -
          RatRef-12 Expression BeadChip ILMN_58552 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline