Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01225236_m1
          See other BCL11A GE Assays ›
          Gene Symbol:
          Bcl11a
          Gene Name:
          B-cell CLL/lymphoma 11A (zinc finger protein)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 108826715 - 108921197 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01225236_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bcl11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191683.1 NP_001178612.1 3 - 4 725 62
          XM_006251579.2 XP_006251641.2 3 - 4 511 62
          XM_006251587.2 XP_006251649.1 3 - 4 646 62
          XM_006251588.2 XP_006251650.1 3 - 4 947 62
          XM_006251591.2 XP_006251653.1 2 - 3 2370 62
          XM_006251594.2 XP_006251656.1 3 - 4 934 62
          XM_008770430.1 XP_008768652.1 3 - 4 947 62
          XM_008770431.1 XP_008768653.1 3 - 4 802 62
          GenBank mRNA BC086607.1 - 3 - 4 834 62
          BC158689.1 - 3 - 4 638 8375842 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400001157.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          negative regulation of gene expression
          positive regulation of neuron projection development
          negative regulation of neuron projection development
          protein sumoylation
          B cell differentiation
          T cell differentiation
          negative regulation of collateral sprouting
          positive regulation of collateral sprouting
          regulation of dendrite development
          negative regulation of nucleic acid-templated transcription
          negative regulation of dendrite development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription corepressor activity
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105903 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105903 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105903 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P414778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58764 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10774604 - -
          Affymetrix Rat Gene 1.0 ST Array 10774597 - -
          Affymetrix Rat Gene 1.0 ST Array 10774601 - -
          Affymetrix Rat Gene 1.0 ST Array 10774600 - -
          Affymetrix Rat Gene 1.0 ST Array 10774598 - -
          Affymetrix Rat Gene 1.0 ST Array 10774599 - -
          Affymetrix Rat Gene 1.1 ST Array 10774596 - -
          Rat Genome 230 2.0 Array 1383506_at - -
          Rat Genome 230 2.0 Array 1383506_PM_at - -
          Affymetrix HT Rat Focus Array 1383506_PM_at - -
          Affymetrix HT Rat Focus Array 1383506_at - -
          Affymetrix Rat Clariom S Assay PSR1400008411.rn.1 TC1400001157.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008409.rn.1 TC1400001157.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008413.rn.1 TC1400001157.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008414.rn.1 TC1400001157.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008412.rn.1 TC1400001157.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400008415.rn.1 TC1400001157.rn.1 RTA TC
          Rat Expression Set 230 1383506_at - -
          Rat Expression Set 230 1383506_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10774596 - -
          Rat Genome U34 Set rc_AA818595_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365365 - -
          RatRef-12 Expression BeadChip ILMN_60640 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline