Search
Search
| Gene Symbol: Traip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109004.2 | NP_001102474.1 | 1 - 2 | 179 | 62 | |
| XM_008766558.1 | XP_008764780.1 | 1 - 2 | 788 | 62 | ||
| GenBank mRNA | BC162006.1 | - | 1 - 2 | 179 | 9089897 | 62 |
Set Membership: |
|