Search
Search
| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022690.2 | NP_073181.1 | 4 - 5 | 734 | 83 | |
| GenBank mRNA | AF099093.1 | - | 4 - 5 | 701 | 83 | |
| BC086980.1 | - | 4 - 5 | 734 | 7192141 | 83 | |
Set Membership: |
|