Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01251028_m1
          See other CBFB GE Assays ›
          Gene Symbol:
          Cbfb
          Gene Name:
          core-binding factor, beta subunit
          Gene Aliases:
          Pebp2
          Chromosome Location:
          Chr.19: 37127508 - 37171075 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01251028_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cbfb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013191.1 NP_001013209.1 4 - 5 679 60
          XM_006255502.2 XP_006255564.1 4 - 5 673 60
          GenBank mRNA BC081946.1 - 4 - 5 679 7121452 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000467.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cbfb

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          biological_process
          lymphocyte differentiation
          myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          definitive hemopoiesis
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P637792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638758 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28488 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P637792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P637792 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P637792 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P637792 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P459533 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701026859 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700626639 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35854 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24514 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92751 - -
          Agilent Rat cDNA Microarray (G4105A) 701026859 - -
          Agilent Rat cDNA Microarray (G4105A) 700626639 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807132 - -
          Affymetrix Rat Gene 1.0 ST Array 10807134 - -
          Affymetrix Rat Gene 1.0 ST Array 10807139 - -
          Affymetrix Rat Gene 1.0 ST Array 10807138 - -
          Affymetrix Rat Gene 1.0 ST Array 10807133 - -
          Affymetrix Rat Gene 1.0 ST Array 10807135 - -
          Affymetrix Rat Gene 1.0 ST Array 10807136 - -
          Affymetrix Rat Gene 1.0 ST Array 10807137 - -
          Affymetrix Rat Gene 1.1 ST Array 10807131 - -
          Rat Genome 230 2.0 Array 1399033_at - -
          Rat Genome 230 2.0 Array 1395821_at - -
          Rat Genome 230 2.0 Array 1399033_PM_at - -
          Rat Genome 230 2.0 Array 1395821_PM_at - -
          Affymetrix HT Rat Focus Array 1395821_at - -
          Affymetrix HT Rat Focus Array 1399033_at - -
          Affymetrix HT Rat Focus Array 1395821_PM_at - -
          Affymetrix HT Rat Focus Array 1399033_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900003710.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003702.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003705.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003707.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003709.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003704.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003706.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003703.rn.1 TC1900000467.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003708.rn.1 TC1900000467.rn.1 RTA TC
          Rat Expression Set 230 1399033_at - -
          Rat Expression Set 230 1399033_PM_at - -
          Rat Expression Set 230 1395821_PM_at - -
          Rat Expression Set 230 1395821_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807131 - -
          Rat Genome U34 Set AF087437UTR#1_s_at - -
          Rat Genome U34 Set rc_AA945869_at - -
          Rat Genome U34 Set rc_AA997973_at - -
          Rat Genome U34 Set rc_AA955334_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21887404 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline