Search
Search
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107403.1 | NP_001100873.1 | 1 - 2 | 51 | 70 | |
| XM_008771946.1 | XP_008770168.1 | 1 - 2 | 179 | 70 | ||
| XM_008771947.1 | XP_008770169.1 | 1 - 2 | 179 | 70 | ||
| XM_008771948.1 | XP_008770170.1 | 1 - 2 | 157 | 70 | ||
| XM_008771949.1 | XP_008770171.1 | 1 - 2 | 179 | 70 | ||
Set Membership: |
|