Search
Search

LOC100910342| Gene Symbol: LOC100910342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749143.3 | XP_003749191.1 | 3 - 4 | 531 | 80 | |
| XM_006231621.2 | XP_006231683.1 | 2 - 3 | 224 | 80 | ||
| XM_008760553.1 | XP_008758775.1 | 4 - 5 | 714 | 80 | ||
| XM_008760554.1 | XP_008758776.1 | 4 - 5 | 688 | 80 | ||
| GenBank mRNA | FQ215762.1 | - | 4 - 5 | 393 | 80 | |
Set Membership: |
|