Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01269940_m1
          See other FGFR2 GE Assays ›
          Gene Symbol:
          Fgfr2
          Gene Name:
          fibroblast growth factor receptor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 200590951 - 200696929 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01269940_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgfr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109892.1 NP_001103362.1 10 - 11 2090 87
          NM_001109893.1 NP_001103363.1 9 - 10 1826 87
          NM_001109894.1 NP_001103364.1 9 - 10 1823 87
          NM_001109895.1 NP_001103365.1 8 - 9 1748 87
          NM_001109896.1 NP_001103366.1 8 - 9 1745 87
          NM_012712.1 NP_036844.1 10 - 11 2093 87
          XM_006230389.2 XP_006230451.1 10 - 11 2088 87
          XM_006230390.2 XP_006230452.1 10 - 11 2085 87
          XM_006230392.2 XP_006230454.1 10 - 11 2094 87
          XM_006230393.2 XP_006230455.1 8 - 9 1740 87
          XM_008759861.1 XP_008758083.1 9 - 10 4250 87
          GenBank mRNA L19109.1 - 2 - 3 227 87
          L19110.1 - 2 - 3 231 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006749.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          ureteric bud development
          in utero embryonic development
          epithelial to mesenchymal transition
          positive regulation of mesenchymal cell proliferation
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          cell-cell signaling
          axonogenesis
          neuromuscular junction development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          regulation of smoothened signaling pathway
          post-embryonic development
          embryonic pattern specification
          organ morphogenesis
          regulation of cell fate commitment
          positive regulation of phospholipase activity
          morphogenesis of embryonic epithelium
          peptidyl-tyrosine phosphorylation
          orbitofrontal cortex development
          ventricular zone neuroblast division
          pyramidal neuron development
          gland morphogenesis
          positive regulation of Wnt signaling pathway
          bone mineralization
          lung development
          epithelial cell differentiation
          midbrain development
          otic vesicle formation
          hair follicle morphogenesis
          lacrimal gland development
          multicellular organism growth
          organ growth
          fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
          fibroblast growth factor receptor signaling pathway involved in hemopoiesis
          fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          regulation of multicellular organism growth
          regulation of fibroblast growth factor receptor signaling pathway
          regulation of cell proliferation
          inner ear morphogenesis
          odontogenesis
          positive regulation of MAPK cascade
          cell fate commitment
          positive regulation of cell cycle
          negative regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          lung alveolus development
          mesodermal cell differentiation
          synaptic vesicle transport
          embryonic digestive tract morphogenesis
          embryonic organ morphogenesis
          digestive tract development
          embryonic organ development
          reproductive structure development
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          epidermis morphogenesis
          branching morphogenesis of a nerve
          mesenchymal cell differentiation
          epithelial cell proliferation
          regulation of epithelial cell proliferation
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          regulation of smooth muscle cell differentiation
          positive regulation of cell division
          ventricular cardiac muscle tissue morphogenesis
          positive regulation of cardiac muscle cell proliferation
          limb bud formation
          bone development
          bone morphogenesis
          coronal suture morphogenesis
          epithelial tube branching involved in lung morphogenesis
          branching involved in prostate gland morphogenesis
          branching involved in salivary gland morphogenesis
          bud elongation involved in lung branching
          lung lobe morphogenesis
          lung-associated mesenchyme development
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          prostate gland morphogenesis
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          lateral sprouting from an epithelium
          mammary gland bud formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          branch elongation involved in salivary gland morphogenesis
          branching involved in labyrinthine layer morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          regulation of morphogenesis of a branching structure
          mesenchymal cell differentiation involved in lung development
          mesenchymal cell proliferation involved in lung development
          endodermal digestive tract morphogenesis
          lens fiber cell development
          regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          fibroblast growth factor-activated receptor activity
          ATP binding
          fibroblast growth factor binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10225 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P064028 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P113700 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P454429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156995 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122589 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132209 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454429 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064028 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P113700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454429 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P064028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10225 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P454429 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P064028 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P113700 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16199 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139894 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454429 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10225 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701977327 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90384 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13586 - -
          Agilent Rat cDNA Microarray (G4105A) 701977327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726188 - -
          Affymetrix Rat Gene 1.0 ST Array 10726185 - -
          Affymetrix Rat Gene 1.0 ST Array 10726179 - -
          Affymetrix Rat Gene 1.0 ST Array 10726176 - -
          Affymetrix Rat Gene 1.0 ST Array 10726182 - -
          Affymetrix Rat Gene 1.0 ST Array 10726178 - -
          Affymetrix Rat Gene 1.0 ST Array 10726183 - -
          Affymetrix Rat Gene 1.0 ST Array 10726190 - -
          Affymetrix Rat Gene 1.0 ST Array 10726192 - -
          Affymetrix Rat Gene 1.0 ST Array 10726173 - -
          Affymetrix Rat Gene 1.0 ST Array 10726187 - -
          Affymetrix Rat Gene 1.0 ST Array 10726189 - -
          Affymetrix Rat Gene 1.0 ST Array 10726174 - -
          Affymetrix Rat Gene 1.0 ST Array 10726177 - -
          Affymetrix Rat Gene 1.0 ST Array 10726186 - -
          Affymetrix Rat Gene 1.0 ST Array 10726184 - -
          Affymetrix Rat Gene 1.0 ST Array 10726180 - -
          Affymetrix Rat Gene 1.0 ST Array 10726181 - -
          Affymetrix Rat Gene 1.0 ST Array 10726191 - -
          Affymetrix Rat Gene 1.0 ST Array 10726175 - -
          Affymetrix Rat Gene 1.1 ST Array 10726172 - -
          Rat Genome 230 2.0 Array 1373829_at - -
          Rat Genome 230 2.0 Array 1373829_PM_at - -
          Rat Genome 230 2.0 Array 1370630_PM_a_at - -
          Rat Genome 230 2.0 Array 1388168_a_at - -
          Rat Genome 230 2.0 Array 1375129_PM_at - -
          Rat Genome 230 2.0 Array 1371179_PM_a_at - -
          Rat Genome 230 2.0 Array 1375129_at - -
          Rat Genome 230 2.0 Array 1371179_a_at - -
          Rat Genome 230 2.0 Array 1388168_PM_a_at - -
          Rat Genome 230 2.0 Array 1370630_a_at - -
          Affymetrix HT Rat Focus Array 1375129_at - -
          Affymetrix HT Rat Focus Array 1370630_a_at - -
          Affymetrix HT Rat Focus Array 1371179_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371179_a_at - -
          Affymetrix HT Rat Focus Array 1375129_PM_at - -
          Affymetrix HT Rat Focus Array 1370630_PM_a_at - -
          Affymetrix HT Rat Focus Array 1388168_PM_a_at - -
          Affymetrix HT Rat Focus Array 1373829_PM_at - -
          Affymetrix HT Rat Focus Array 1373829_at - -
          Affymetrix HT Rat Focus Array 1388168_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100053294.rn.1 TC0100006749.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100053295.rn.1 TC0100006749.rn.1 RTA TC
          Rat Expression Set 230 1371179_a_at - -
          Rat Expression Set 230 1370630_a_at - -
          Rat Expression Set 230 1373829_at - -
          Rat Expression Set 230 1370630_PM_a_at - -
          Rat Expression Set 230 1373829_PM_at - -
          Rat Expression Set 230 1388168_a_at - -
          Rat Expression Set 230 1371179_PM_a_at - -
          Rat Expression Set 230 1375129_at - -
          Rat Expression Set 230 1375129_PM_at - -
          Rat Expression Set 230 1388168_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726172 - -
          Rat Genome U34 Set L19107_s_at - -
          Rat Genome U34 Set L19112_at - -
          Rat Genome U34 Set Z35138cds_s_at - -
          Rat Genome U34 Set L19112_g_at - -
          Rat Genome U34 Set rc_AA997521_at - -
          Rat Genome U34 Set rc_AI229178_at - -
          Rat Genome U34 Set rc_AI111547_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21846960 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371701 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline