Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01274782_g1
          See other RGD1311703 GE Assays ›
          Gene Symbol:
          RGD1311703
          Gene Name:
          similar to sid2057p
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 185557633 - 185569190 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01274782_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1311703
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006230086.2 XP_006230148.1 4 - 5 465 97
          GenBank mRNA BC089924.1 - 4 - 5 430 7307309 97
          FM053223.1 - 4 - 5 469 97
          FQ225875.1 - 4 - 5 465 97
          FQ227306.1 - 4 - 5 465 97

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC089924.1 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P996917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145417 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57945 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P996917 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P996917 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P996917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70860 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710130 - -
          Affymetrix Rat Gene 1.0 ST Array 10710133 - -
          Affymetrix Rat Gene 1.0 ST Array 10710131 - -
          Affymetrix Rat Gene 1.0 ST Array 10710134 - -
          Affymetrix Rat Gene 1.0 ST Array 10710132 - -
          Affymetrix Rat Gene 1.1 ST Array 10710129 - -
          Rat Genome 230 2.0 Array 1389575_at - -
          Rat Genome 230 2.0 Array 1389575_PM_at - -
          Affymetrix HT Rat Focus Array 1389575_PM_at - -
          Affymetrix HT Rat Focus Array 1389575_at - -
          Affymetrix Rat Clariom S Assay PSR0100050761.rn.1 TC0100006472.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100050759.rn.1 TC0100006472.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100050760.rn.1 TC0100006472.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100050762.rn.1 TC0100006472.rn.1 RTA TC
          Rat Expression Set 230 1389575_at - -
          Rat Expression Set 230 1389575_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710129 - -
          Rat Genome U34 Set rc_AI233370_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21964442 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline