Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01279391_m1
          See other WEE1 GE Assays ›
          Gene Symbol:
          Wee1
          Gene Name:
          WEE1 G2 checkpoint kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 174765230 - 174790863 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01279391_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wee1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012742.1 NP_001012760.1 10 - 11 1848 87
          XM_006229974.2 XP_006230036.1 10 - 11 4578 87
          GenBank mRNA BC090346.1 - 10 - 11 1848 7306767 87
          D31838.1 - 10 - 11 2047 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002354.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wee1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          protein phosphorylation
          mitotic nuclear division
          peptidyl-tyrosine phosphorylation
          establishment of cell polarity
          neuron projection morphogenesis
          cell division
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P487112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158713 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487112 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P487112 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P487112 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409975 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487112 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701332334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18918 - -
          Agilent Rat cDNA Microarray (G4105A) 701332334 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709846 - -
          Affymetrix Rat Gene 1.0 ST Array 10709849 - -
          Affymetrix Rat Gene 1.0 ST Array 10709848 - -
          Affymetrix Rat Gene 1.0 ST Array 10709857 - -
          Affymetrix Rat Gene 1.0 ST Array 10709852 - -
          Affymetrix Rat Gene 1.0 ST Array 10709853 - -
          Affymetrix Rat Gene 1.0 ST Array 10709847 - -
          Affymetrix Rat Gene 1.0 ST Array 10709855 - -
          Affymetrix Rat Gene 1.0 ST Array 10709851 - -
          Affymetrix Rat Gene 1.0 ST Array 10709854 - -
          Affymetrix Rat Gene 1.0 ST Array 10709856 - -
          Affymetrix Rat Gene 1.0 ST Array 10709850 - -
          Affymetrix Rat Gene 1.1 ST Array 10709844 - -
          Rat Genome 230 2.0 Array 1370663_PM_at - -
          Rat Genome 230 2.0 Array 1370663_at - -
          Rat Genome 230 2.0 Array 1397409_PM_s_at - -
          Rat Genome 230 2.0 Array 1397409_s_at - -
          Affymetrix HT Rat Focus Array 1397409_s_at - -
          Affymetrix HT Rat Focus Array 1397409_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370663_at - -
          Affymetrix HT Rat Focus Array 1370663_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100017852.rn.1 TC0100002354.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100017851.rn.1 TC0100002354.rn.1 RTA TC
          Rat Expression Set 230 1370663_at - -
          Rat Expression Set 230 1370663_PM_at - -
          Rat Expression Set 230 1397409_s_at - -
          Rat Expression Set 230 1397409_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709844 - -
          Rat Genome U34 Set D31838_at - -
          Rat Genome U34 Set D31838_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21720953 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-vdfpx:80/100.66.78.86:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0