Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01286286_g1
          See other SOX2 GE Assays ›
          Gene Symbol:
          Sox2
          Gene Name:
          SRY (sex determining region Y)-box 2
          Gene Aliases:
          RGD1565646
          Chromosome Location:
          Chr.2: 121062030 - 121203880 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01286286_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109181.1 NP_001102651.1 1 - 2 1535 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000979.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          HMG box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          osteoblast differentiation
          eye development
          cell fate specification
          endodermal cell fate specification
          positive regulation of neuroblast proliferation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell cycle arrest
          sensory perception of sound
          response to wounding
          response to organic substance
          regulation of gene expression
          stem cell population maintenance
          forebrain neuron differentiation
          pituitary gland development
          adenohypophysis development
          cerebral cortex development
          positive regulation of cell-cell adhesion
          negative regulation of Wnt signaling pathway
          male genitalia development
          positive regulation of epithelial cell differentiation
          forebrain development
          olfactory placode formation
          response to retinoic acid
          somatic stem cell population maintenance
          tissue regeneration
          inner ear morphogenesis
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          tongue development
          cell fate commitment
          negative regulation of cell differentiation
          positive regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          pigment biosynthetic process
          lung alveolus development
          embryonic organ development
          anatomical structure formation involved in morphogenesis
          neuron fate commitment
          inner ear development
          diencephalon morphogenesis
          stem cell differentiation
          negative regulation of epithelial cell proliferation
          regulation of neurogenesis
          detection of mechanical stimulus involved in sensory perception of sound
          detection of mechanical stimulus involved in equilibrioception
          retina morphogenesis in camera-type eye
          lens induction in camera-type eye
          epithelial tube branching involved in lung morphogenesis
          response to growth factor
          negative regulation of canonical Wnt signaling pathway
          neuronal stem cell population maintenance
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          molecular_function
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          miRNA binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P506024 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P778330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125378 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P778330 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P506024 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P778330 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P506024 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P778330 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P506024 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814848 - -
          Affymetrix Rat Gene 1.1 ST Array 10814847 - -
          Rat Genome 230 2.0 Array 1375153_at - -
          Rat Genome 230 2.0 Array 1379942_PM_at - -
          Rat Genome 230 2.0 Array 1389826_PM_at - -
          Rat Genome 230 2.0 Array 1381874_PM_at - -
          Rat Genome 230 2.0 Array 1381874_at - -
          Rat Genome 230 2.0 Array 1375153_PM_at - -
          Rat Genome 230 2.0 Array 1389826_at - -
          Rat Genome 230 2.0 Array 1379942_at - -
          Affymetrix HT Rat Focus Array 1375153_at - -
          Affymetrix HT Rat Focus Array 1381874_PM_at - -
          Affymetrix HT Rat Focus Array 1389826_at - -
          Affymetrix HT Rat Focus Array 1381874_at - -
          Affymetrix HT Rat Focus Array 1379942_PM_at - -
          Affymetrix HT Rat Focus Array 1379942_at - -
          Affymetrix HT Rat Focus Array 1375153_PM_at - -
          Affymetrix HT Rat Focus Array 1389826_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200006008.rn.1 TC0200000979.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200006006.rn.1 TC0200000979.rn.1 RTA TC
          Rat Expression Set 230 1389826_at - -
          Rat Expression Set 230 1375153_PM_at - -
          Rat Expression Set 230 1389826_PM_at - -
          Rat Expression Set 230 1375153_at - -
          Rat Expression Set 230 1379942_PM_at - -
          Rat Expression Set 230 1381874_at - -
          Rat Expression Set 230 1381874_PM_at - -
          Rat Expression Set 230 1379942_at - -
          Affymetrix Rat Gene 1.0 ST Array 10814847 - -
          Rat Genome U34 Set rc_AI102567_at - -
          Rat Genome U34 Set rc_AA858564_at - -
          Rat Genome U34 Set rc_AI111954_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21982763 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline