Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01297610_m1
          See other NKAP GE Assays ›
          Gene Symbol:
          Nkap
          Gene Name:
          NFKB activating protein
          Gene Aliases:
          2610020o08rik
          Chromosome Location:
          Chr.X: 123751196 - 123770595 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01297610_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024872.1 NP_001020043.1 2 - 3 558 87
          GenBank mRNA BC098011.1 - 2 - 3 558 7454308 87
          FQ234084.1 - 2 - 3 594 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001066.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkap

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          Notch signaling pathway
          stem cell population maintenance
          hemopoiesis
          granulocyte differentiation
          T cell differentiation in thymus
          negative regulation of transcription, DNA-templated
          positive regulation of alpha-beta T cell differentiation
          hematopoietic stem cell proliferation
          chromatin binding
          chromatin DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1011018 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166587 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112610 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1011018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1011018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1011018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93276 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28263 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63024 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936290 - -
          Affymetrix Rat Gene 1.0 ST Array 10936295 - -
          Affymetrix Rat Gene 1.0 ST Array 10936298 - -
          Affymetrix Rat Gene 1.0 ST Array 10936299 - -
          Affymetrix Rat Gene 1.0 ST Array 10936296 - -
          Affymetrix Rat Gene 1.0 ST Array 10936297 - -
          Affymetrix Rat Gene 1.0 ST Array 10936292 - -
          Affymetrix Rat Gene 1.0 ST Array 10936294 - -
          Affymetrix Rat Gene 1.0 ST Array 10936291 - -
          Affymetrix Rat Gene 1.0 ST Array 10936293 - -
          Affymetrix Rat Gene 1.1 ST Array 10936289 - -
          Rat Genome 230 2.0 Array 1384761_at - -
          Rat Genome 230 2.0 Array 1382109_at - -
          Rat Genome 230 2.0 Array 1385495_PM_at - -
          Rat Genome 230 2.0 Array 1385495_at - -
          Rat Genome 230 2.0 Array 1384761_PM_at - -
          Rat Genome 230 2.0 Array 1382109_PM_at - -
          Affymetrix HT Rat Focus Array 1382109_at - -
          Affymetrix HT Rat Focus Array 1385495_at - -
          Affymetrix HT Rat Focus Array 1382109_PM_at - -
          Affymetrix HT Rat Focus Array 1385495_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006594.rn.1 TC0X00001066.rn.1 RTA TC
          Rat Expression Set 230 1385495_at - -
          Rat Expression Set 230 1382109_at - -
          Rat Expression Set 230 1385495_PM_at - -
          Rat Expression Set 230 1382109_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936289 - -
          Rat Genome U34 Set rc_AI030080_at - -
          Rat Genome U34 Set rc_AI012804_at - -
          Rat Genome U34 Set rc_AI172058_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369537 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline