Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01299126_m1
See other CACNA1G GE Assays ›
Gene Symbol:
Cacna1g
Gene Name:
calcium channel, voltage-dependent, T type, alpha 1G subunit
Gene Aliases:
-
Chromosome Location:
Chr.10: 82129071 - 82197863 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01299126_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Cacna1g
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_031601.3
NP_113789.2
22 - 23
4838
68
XM_008767985.1
XP_008766207.1
23 - 24
4925
68
XM_008767986.1
XP_008766208.1
23 - 24
4925
68
XM_008767987.1
XP_008766209.1
23 - 24
4925
68
XM_008767988.1
XP_008766210.1
22 - 23
4856
68
XM_008767989.1
XP_008766211.1
23 - 24
4925
68
XM_008767990.1
XP_008766212.1
23 - 24
4925
68
XM_008767991.1
XP_008766213.1
23 - 24
4925
68
XM_008767992.1
XP_008766214.1
23 - 24
4925
68
XM_008767993.1
XP_008766215.1
22 - 23
4856
68
XM_008767994.1
XP_008766216.1
23 - 24
4925
68
XM_008767995.1
XP_008766217.1
23 - 24
4925
68
XM_008767996.1
XP_008766218.1
23 - 24
4925
68
XM_008767997.1
XP_008766219.1
22 - 23
4856
68
XM_008767998.1
XP_008766220.1
23 - 24
4925
68
XM_008767999.1
XP_008766221.1
22 - 23
4856
68
XM_008768000.1
XP_008766222.1
22 - 23
4805
68
XM_008768001.1
XP_008766223.1
22 - 23
4856
68
GenBank mRNA
AF027984.1
-
22 - 23
4838
68
AF125161.1
-
23 - 24
4582
68
AF290212.2
-
22 - 23
4469
68
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC1000003589.rn.1
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Ion Channel
Recommended Assay
Rattus norvegicus
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Cacna1g
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
action potential
regulation of heart rate
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
response to nickel cation
artery smooth muscle contraction
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of calcium ion-dependent exocytosis
regulation of calcium ion transport
regulation of atrial cardiac muscle cell membrane depolarization
calcium ion transport into cytosol
calcium ion import
calcium ion transmembrane transport
cellular response to dexamethasone stimulus
membrane depolarization during action potential
voltage-gated calcium channel activity
low voltage-gated calcium channel activity
scaffold protein binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Rat Clariom S Assay
PSR1000031979.rn.1
TC1000003589.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000031981.rn.1
TC1000003589.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR1000031980.rn.1
TC1000003589.rn.1
RTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline