Search
Search
| Gene Symbol: Pfas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105791.1 | NP_001099261.1 | 6 - 7 | 748 | 63 | |
| XM_006246610.2 | XP_006246672.1 | 7 - 8 | 1106 | 63 | ||
| XM_006246611.2 | XP_006246673.1 | 6 - 7 | 929 | 63 | ||
| XM_006246612.2 | XP_006246674.1 | 5 - 6 | 638 | 63 | ||
Set Membership: |
|